Skip to Content
MilliporeSigma

Skip To

1602502

USP

Retinyl palmitate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Vitamin A palmitate, all−trans−Retinol palmitate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C36H60O2
CAS Number:
Molecular Weight:
524.86
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1917366
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

retinyl compounds

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1=C(/C=C/C(C)=C/C=C/C(C)=C/COC(CCCCCCCCCCCCCCC)=O)C(C)(C)CCC1

InChI

1S/C36H60O2/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-25-35(37)38-30-28-32(3)23-20-22-31(2)26-27-34-33(4)24-21-29-36(34,5)6/h20,22-23,26-28H,7-19,21,24-25,29-30H2,1-6H3/b23-20+,27-26+,31-22+,32-28+

InChI key

VYGQUTWHTHXGQB-FFHKNEKCSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Review: Vitamin A metabolism.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
R3375PHR1235R1512
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

-

grade

certified reference material, pharmaceutical secondary standard

grade

-

format

neat

format

-

format

neat

format

-

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

pharmaceutical (small molecule)

application(s)

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

API family

retinyl compounds

API family

-

API family

retinyl compounds

API family

-


Still not finding the right product?

Explore all of our products under Retinyl palmitate


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 2

flash_point_f

381.2 °F

flash_point_c

194.00 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Vitamin A
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(2), 4632-4632 (2019)
Vitamin A Assay
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 38(6), 6801-6801 (2014)
Shaikh Meshbahuddin Ahmad et al.
Contemporary clinical trials, 39(2), 269-279 (2014-10-02)
In recent years, neonatal vitamin A supplementation is considered as an essential infant-survival intervention but the evidence is not conclusive. This randomized controlled clinical trial was conducted to evaluate the effect of vitamin A on immune competence in early infancy.



Global Trade Item Number

SKUGTIN
1602502-4X50MG04061838745361

Questions

Reviews

No rating value

Active Filters