Skip to Content
MilliporeSigma

1716002

USP

Retinyl acetate

United States Pharmacopeia (USP) Reference Standard, solution of retinyl acetate (vitamin A) in peanut oil

Synonym(s):

Retinyl acetate solution, Vitamin A acetate solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H32O2
CAS Number:
Molecular Weight:
328.49
UNSPSC Code:
41116107
PubChem Substance ID:
EC Number:
204-844-2
NACRES:
NA.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

retinyl compounds

form

solution

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-8°C

SMILES string

O(C\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(CCCC1(C)C)C)C(=O)C

InChI

1S/C22H32O2/c1-17(9-7-10-18(2)14-16-24-20(4)23)12-13-21-19(3)11-8-15-22(21,5)6/h7,9-10,12-14H,8,11,15-16H2,1-6H3/b10-7+,13-12+,17-9+,18-14+

InChI key

QGNJRVVDBSJHIZ-QHLGVNSISA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Retinyl acetate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Oil- and Water-Soluble Vitamins Capsules
  • Oil- and Water-Soluble Vitamins Oral Solution
  • Oil- and Water-Soluble Vitamins Tablets
  • Oil- and Water-Soluble Vitamins with Minerals Capsules
  • Oil- and Water-Soluble Vitamins with Minerals Chewable Gels
  • Oil- and Water-Soluble Vitamins with Minerals Oral Solution
  • Oil- and Water-Soluble Vitamins with Minerals Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Retinyl acetate


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Aquatic Chronic 4 - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library