Skip to Content
MilliporeSigma

Skip To

1546489

USP

Polyethylene glycol 1000

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Poly(ethylene glycol), PEG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

polyethylene glycol

form

solid

mol wt

Mw 950-1050

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

General description

Polyethylene glycol 1000 (PEG 1000) is a water-soluble polymer used widely in biologics and pharmaceutical formulations. As a carbohydrate, it plays a vital role in enhancing the stability and solubility of biologic drugs. PEG 1000 is particularly significant in the development of sustained-release formulations and protein conjugation, as it reduces immunogenicity and extends the half-life of biologic therapeutics. Its non-toxic, biocompatible properties make it essential in the production of injectable biologics, improving their overall efficacy and safety profile.

The USP biologics carbohydrates category includes a variety of carbohydrate-based substances that are essential in the development and manufacturing of therapeutic products. These carbohydrates play crucial roles in biological processes and are often utilized as excipients, stabilizers, or active ingredients in pharmaceuticals. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of carbohydrate therapeutics throughout their lifecycle.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Polyethylene glycol 1000 USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Polyoxyl 15 Hydroxystearate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
This product is part of USP Biologics Program.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pha
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
154658015464671546423
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

form

solid

form

-

form

-

form

-

format

neat

format

neat

format

neat

format

neat

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

mol wt

Mw 950-1050

mol wt

Mw 5400-6600

mol wt

Mw 570-630

mol wt

Mw 285-315


Still not finding the right product?

Explore all of our products under Polyethylene glycol 1000


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

281.5 °F - closed cup

flash_point_c

138.6 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1546489-1G04061833860953

Questions

Reviews

No rating value

Active Filters