Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1546547

USP

Polyethylene glycol 3350

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Poly(ethylene glycol), PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

polyethylene glycol

mol wt

Mw 3015-3685

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

For further information and support please go to the website of the issuing Pharmacopoeia.

Polyethylene Glycol 3350 is an addition polymer of ethylene oxide and water, represented by the formula H(OCH2CH2)nOH, in which n represents the average number of oxyethylene groups. The apparent weight-average molecular weight is 3015–3685 g/mol (Da).

Application

Polyethylene glycol 3350 USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Polyethylene Glycol 3350 and Electrolytes for Oral Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyethylene Glycol 3350
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(4), 3584-3584 (2020)
Polyethylene Glycol 3350 and Electrolytes for Oral Solution
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 33(6), 3587-3587 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service