Skip to Content
MilliporeSigma

Skip To

1133547

USP

Choline chloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(2-Hydroxyethyl)trimethylammonium chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)3N(Cl)CH2CH2OH
CAS Number:
Molecular Weight:
139.62
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3563126
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

choline chloride

manufacturer/tradename

USP

mp

302-305 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Cl-].C[N+](C)(C)CCO

InChI

1S/C5H14NO.ClH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H/q+1;/p-1

InChI key

SGMZJAMFUVOLNK-UHFFFAOYSA-M

General description

Choline acetyltransferase substrate
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
5.00117W450001C7527
grade

pharmaceutical primary standard

grade

-

grade

Halal, Kosher

grade

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
solid formulation

application(s)

flavors and fragrances

application(s)

-

format

neat

format

-

format

-

format

-

mp

302-305 °C (dec.) (lit.)

mp

-

mp

302-305 °C (dec.) (lit.)

mp

302-305 °C (dec.) (lit.)

API family

choline chloride

API family

-

API family

-

API family

-


Still not finding the right product?

Explore all of our products under Choline chloride


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters