Skip to Content
MilliporeSigma
Choline chloride Pharmaceutical Secondary Standard; Certified Reference Material

Choline chloride

Choline chloride Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
(2-Hydroxyethyl)trimethylammonium chloride, Choline chloride
Linear Formula:
(CH3)3N(Cl)CH2CH2OH
CAS Number:
Molecular Weight:
139.62
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique