Skip to Content
MilliporeSigma

Skip To

1067001

USP

Betamethasone acetate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Betamethasone 21-acetate, 9α-Fluoro-16β-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione 21-acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C24H31FO6
CAS Number:
Molecular Weight:
434.50
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

betamethasone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(C)=O

InChI

1S/C24H31FO6/c1-13-9-18-17-6-5-15-10-16(27)7-8-21(15,3)23(17,25)19(28)11-22(18,4)24(13,30)20(29)12-31-14(2)26/h7-8,10,13,17-19,28,30H,5-6,9,11-12H2,1-4H3/t13-,17-,18-,19-,21-,22-,23-,24-/m0/s1

InChI key

AKUJBENLRBOFTD-QZIXMDIESA-N

Gene Information

human ... NR3C1(2908)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Betamethasone acetate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Betamethasone Sodium Phosphate and Betamethasone Acetate Injectable Suspension
  • Gentamicin Sulfate and Betamethasone Acetate Ophthalmic Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1779B10300001556008
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

EDQM

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

API family

betamethasone

API family

betamethasone

API family

betamethasone

API family

prednisolone

Gene Information

human ... NR3C1(2908)

Gene Information

human ... NR3C1(2908)

Gene Information

human ... NR3C1(2908)

Gene Information

human ... NR3C1(2908)


Still not finding the right product?

Explore all of our products under Betamethasone acetate


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Repr. 1B - STOT RE 2

target_organs

Liver,Kidney,Endocrine system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Andrejus Korolkovas
ESSENTIALS OF MEDICINAL CHEMISTRY, 1044-1044 (2008)
Cody C Wyles et al.
Clinical orthopaedics and related research, 473(3), 1155-1164 (2014-09-05)
Corticosteroids are a common, short-term, local antiinflammatory and analgesic for treating patients with musculoskeletal disorders. Studies have shown the deleterious effects of corticosteroids on chondrocytes, suggesting a potentiation of degenerative joint disease. Mesenchymal stem cells (MSCs) are the direct progenitors



Global Trade Item Number

SKUGTIN
1067001-500MG04061833849231

Questions

Reviews

No rating value

Active Filters