Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1556008

USP

Prednisolone acetate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Prednisolone 21-acetate, 1,4-Pregnadiene-11β,17α,21-triol-3,20-dione 21-acetate, 11β,17α,21-Trihydroxy-1,4-pregnadiene-3,20-dione 21-acetate, 21-Acetoxy-1,4-pregnadiene-11β,17α-diol-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30O6
CAS Number:
Molecular Weight:
402.48
Beilstein:
3111798
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

prednisolone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)COC(C)=O

InChI

1S/C23H30O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h6,8,10,16-18,20,26,28H,4-5,7,9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1

InChI key

LRJOMUJRLNCICJ-JZYPGELDSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rohit Shetty et al.
Indian journal of ophthalmology, 62(9), 923-926 (2014-11-06)
To study the safety and efficacy of sutureless femtosecond anterior lamellar keratoplasty (FALK) in patients with corneal stromal opacities. Eleven eyes of 11 consecutive patients with corneal stromal opacities involving < 250 μ due to various pathologies were included in
Oscar A Candia et al.
Experimental eye research, 128, 114-116 (2014-10-12)
We have previously shown that tissue plasminogen activator (tPA) injected in the vitreous of sheep, reduced or prevented the elevation of the intraocular pressure (IOP) normally produced by the instillation of 1% prednisolone. We now report the effect of tPA
Rohit Shetty et al.
American journal of ophthalmology, 159(3), 419-425 (2014-12-03)
To evaluate the effect of keratoconus cone location on the change in refractive outcomes, corneal aberrations, and biomechanics after combined topography-guided photorefractive keratectomy (PRK) and collagen cross-linking (CXL). Prospective, comparative case series. Topography-guided PRK was performed followed by accelerated CXL
Antigoni Koukkoulli et al.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie, 253(1), 99-106 (2014-10-12)
To report on long-term outcomes of mitomycin C (MMC)-augmented needle revision of failing deep sclerectomy (DS) blebs. Retrospective database search of all needle revisions with MMC for DS blebs between 2002 and 2008 was conducted. Sixty-six eyes of 66 patients
Salima I Hassanaly et al.
American journal of ophthalmology, 158(2), 270-276 (2014-05-23)
To describe outcomes after Boston Type 1 Keratoprosthesis (KPro) surgery in aniridic eyes. Retrospective, interventional case series. University-based tertiary care institution. Twenty-six aniridic eyes of 19 patients who underwent KPro implantation by a single experienced surgeon (M.H.-D.) between October 27

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service