Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NB.22
UNSPSC Code:
41122102
Manufacturer/tradename:
Diversified Biotech WPNP-1000
Packaging:
pack of 50 ea
Pricing and availability is not currently available.
sterility
non-sterile
packaging
pack of 50 ea
manufacturer/tradename
Diversified Biotech WPNP-1000
size
96 wells
color
none
General description
A note pad that corresponds to the layout of a 96-, 48- or 24-well plate. A fast and easy way to keep accurate records of samples in a microplate. The adhesive backing allows the researcher to affix individual sheets into a notebook. The pad contains (50) 5" × 7" sheets.
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename Diversified Biotech WPNP-1000 | manufacturer/tradename Diversified Biotech WPST-1000 | manufacturer/tradename MC-Media Pad® | manufacturer/tradename Corning 3658 |
| packaging pack of 50 ea | packaging pkg of 1 Stand | packaging pack of 4 × 25 pieces | packaging case of 100, pack of 25 |
| sterility non-sterile | sterility non-sterile | sterility - | sterility non-sterile |
| size 96 wells | size - | size - | size 384 wells |
| color none | color none | color - | color black |
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



