Skip to Content
MilliporeSigma
All Photos(3)

Documents

1.32357

Millipore

MC-Media Pad®

for coliforms, for Escherichia coli

Synonym(s):

E. coli and coliform test

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171504
NACRES:
NA.75

Agency

AOAC® PTM
MicroVal

product line

MC-Media Pad®

form

solid

feature

ready-to-use

manufacturer/tradename

MC-Media Pad®

storage condition

(Store upright)

pH

7.4 ( in H2O)

application(s)

food and beverages
personnel monitoring
surface monitoring
water monitoring

shipped in

ambient

storage temp.

2-25°C

suitability

Escherichia coli
enterovirulent E. coli (EEC) spp.
coliforms
molds
yeasts

General description

The MC MEDIA PAD® offers a convenient solution for rapidly and routinely testing microbial contamination in raw and in-process food and beverage materials, as well as finished products. The MC-Media Pad® is a pre-sterilized, ready-to-use dry culture device, simplifying testing and minimizing the quantity of waste. The media pad consists of a unique adhesive sheet, a test pad coated with medium and water absorption polymer, and a transparent cover film. MC-Media Pad® E. coli and Coliform Count is for simultaneous enumeration of E. coli and the total number of coliforms present in food or beverage samples. The growth medium coats the media pad, and a redox indicator is present for detection. Once inoculated onto the test pad, the liquid sample automatically diffuses through the whole pad by capillary action.

Application

To determine the microbial count in food and beverage products. MC MEDIA PAD® E. coli & Coliform is intended to simultaneously detect and enumerate coliforms and E. coli through special medium composition and specific chromogenic substrates for both β-galactosidase and β-glucuronidase. While improving the workflow, it also ensures accurate and reliable results.

Features and Benefits

  • Save space in fridge and incubator
  • Go green & gain: reduce environmental impact
  • Improve the inventory management with a shelf-life of up to 18 months
  • Comply with regulations
  • No spreading device required
  • Simplified workflow

Analysis Note

If target organisms are present, coliform grow as red-purple/navy color and E. coli colonies on the test pad.

Legal Information

AOAC is a registered trademark of AOAC International
MC MEDIA PAD is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This article will focus on results obtained for MC-Media Pad RAC, MC-Media Pad Coliform, MC-Media Pad E. coli & Coliform & MC-Media Pad Yeast & Mold.

Updates on ISO standards in food microbiology by expert Barbara Gerten from Merck KGaA.

Updates on ISO standards in food microbiology by expert Barbara Gerten from Merck KGaA.

Updates on ISO standards in food microbiology by expert Barbara Gerten from Merck KGaA.

See All

Protocols

Comparison of MC-Media Pad® with different convenience solution products for indicator organism testing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service