Skip to Content
MilliporeSigma

Skip To

SML2663

Selamectin

≥98% (HPLC)

Synonym(s):

PF-2636204, PHA-708778, UK-124114, (5Z)-25-Cyclohexyl-4′-O-de(2,6-dideoxy-3-O-methyl-α-L-arabino-hexopyranosyl)-5-demethoxy-25-de(1-methylpropyl)-22,23-dihydro-5-(hydroxyimino)avermectin A1a

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C43H63NO11
CAS Number:
Molecular Weight:
769.96
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O/N=C1[C@]2([H])[C@]3(O)[C@@](C(O[C@@H]4C[C@]5(O[C@@](C/C=C(C)/[C@@H](O[C@]6([H])C[C@@H]([C@@H](O)[C@H](C)O6)OC)[C@@H](C)/C=C/C=C3\CO2)([H])C4)O[C@H](C7CCCCC7)[C@@H](C)CC5)=O)([H])C=C\1C

InChI

1S/C43H63NO11/c1-24-11-10-14-30-23-50-40-36(44-48)27(4)19-33(43(30,40)47)41(46)52-32-20-31(16-15-25(2)38(24)53-35-21-34(49-6)37(45)28(5)51-35)54-42(22-32)18-17-26(3)39(55-42)29-12-8-7-9-13-29/h10-11,14-15,19,24,26,28-29,31-35,37-40,45,47-48H,7-9,12-13,16-18,20-23H2,1-6H3/b11-10+,25-15+,30-14+,44-36-/t24-,26-,28-,31+,32-,33-,34-,35-,37-,38-,39-,40+,42+,43+/m0/s1

InChI key

AFJYYKSVHJGXSN-XHKIUTQPSA-N

Biochem/physiol Actions

Selamectin is a veterinary parasiticide and anthelminthic. Its mechanism of action is by activating glutamate-gated chloride channels at muscle synapses.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1611729Y000081432476
assay

≥98% (HPLC)

assay

-

assay

-

assay

-

form

powder

form

-

form

-

form

-

storage temp.

−20°C

storage temp.

-

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

-

solubility

-

solubility

-

color

white to beige

color

-

color

-

color

-


Still not finding the right product?

Explore all of our products under Selamectin


hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Questions

Reviews

No rating value

Active Filters