Skip to Content
MilliporeSigma

32476

Selamectin

VETRANAL®, analytical standard

Synonym(s):

(5Z)-25-Cyclohexyl-4′-O-de(2,6-dideoxy-3-O-methyl-α-L-arabino-hexopyranosyl)-5-demethoxy-25-de(1-methylpropyl)-22,23-dihydro-5-(hydroxyimino)avermectin A1a

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C43H63NO11
CAS Number:
Molecular Weight:
769.96
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O/N=C1[C@]2([H])[C@]3(O)[C@@](C(O[C@@H]4C[C@]5(O[C@@](C/C=C(C)/[C@@H](O[C@]6([H])C[C@@H]([C@@H](O)[C@H](C)O6)OC)[C@@H](C)/C=C/C=C3\CO2)([H])C4)O[C@H](C7CCCCC7)[C@@H](C)CC5)=O)([H])C=C\1C

InChI

1S/C43H63NO11/c1-24-11-10-14-30-23-50-40-36(44-48)27(4)19-33(43(30,40)47)41(46)52-32-20-31(16-15-25(2)38(24)53-35-21-34(49-6)37(45)28(5)51-35)54-42(22-32)18-17-26(3)39(55-42)29-12-8-7-9-13-29/h10-11,14-15,19,24,26,28-29,31-35,37-40,45,47-48H,7-9,12-13,16-18,20-23H2,1-6H3/b11-10+,25-15+,30-14+,44-36-/t24-,26-,28-,31+,32-,33-,34-,35-,37-,38-,39-,40+,42+,43+/m0/s1

InChI key

AFJYYKSVHJGXSN-XHKIUTQPSA-N

General description

Selamectin (25-cyclohexyl-25-de(1-methylpropyl)-5-deoxy-22,23-dihydro-5-(hydroxyimino)-avermectin B1 monosaccharide) is a novel endectocide. It is an anti-parasitic agent and has a unique combination of safety and efficacy both in cats and dogs following both topical and oral administration. It prevents ticks and fleas, immature heartworms, ascarids and intestinal hookworms. It can be easily tolerated even at higher dosages than 22,23-dihydroavermectin B1a (DHAVM).

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Selamectin


hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library