Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.25
UNSPSC Code:
41141827
Biological source:
fermentation/recombinant (E. coli strain DH5A)
Form:
liquid
Solubility:
water: soluble at 20 °C
Storage temp.:
2-8°C
Pricing and availability is not currently available.
biological source
fermentation/recombinant (E. coli strain DH5A)
Quality Segment
grade
Molecular Biology
form
liquid
usage
80 uses
solubility
water: soluble at 20 °C
suitability
suitable for electrophoresis (DNA)
storage temp.
2-8°C
General description
Sigma′s PCR 20 bp Low Ladder contains 50 dsDNA fragments from 50 – 1,000 bases, evenly spaced in 20 bp repeats. Five μl of the ladder should be diluted in gel loading buffer and then loaded in a single lane on an agarose gel.
Application
PCR 20 bp Low Ladder has been used as a marker for DNA gel electrophoresis.
Other Notes
Contains 50 bands, 20–1,000 bp in exact 20 bp repeats.
For optimal resolution, the recommended agarose gel concentration is 2.5%.
Sigma′s PCR 20 bp Low Marker is provided in a solution of 10 mM Tris-HCl (pH 8.0), with 1.0 mM EDTA.
1 of 1
This Item | |||
|---|---|---|---|
| grade Molecular Biology | grade Molecular Biology | grade Molecular Biology | grade - |
| biological source fermentation/recombinant (E. coli strain DH5A) | biological source - | biological source synthetic | biological source - |
| Quality Level 100 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| form liquid | form liquid | form liquid | form liquid |
| solubility water: soluble at 20 °C | solubility water: soluble | solubility - | solubility - |
| storage temp. 2-8°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| P1598-25UG | 04061831594997 |
| P1598-40UG | 04061835573783 |



