Skip to Content
MilliporeSigma

Skip To

F6755

Fibrinogen from rat plasma

60-80% protein (≥60% of protein is clottable)

Synonym(s):

Factor I

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-598-6
NACRES:
NA.61
MDL number:
Form:
powder
Biological source:
rat plasma
Mol wt:
α-chain 63.5 kDa, β-chain 56 kDa, γ chain 47 kDa (about 4% carbohydrate content), soluble dimer 340 kDa
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat plasma

Quality Level

form

powder

quality

60-80% protein (≥60% of protein is clottable)

mol wt

α-chain 63.5 kDa, β-chain 56 kDa, γ chain 47 kDa (about 4% carbohydrate content), soluble dimer 340 kDa

concentration

60-80% (biuret)

technique(s)

cell culture | mammalian: suitable, immunoblotting: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

Application

Fibrinogen has been used in studies of haemostatic therapy in surgical and massive trauma patients. These studies have shown that fibrinogen may prove to be more superior in stopping blood loss when compared to using fresh frozen plasma (FFP).
Fibrinogen from rat plasma has been used:
  • in the preparation of growth factor solutions[1]
  • to apply on discontinuous SDS-polyacrylamide gels for the detection of IgG (immunoglobulins) type antibodies by immunoblot analysis
  • to mix with cell suspension to form a clot and retain the cells for avoiding its dispersion during transplantation

Biochem/physiol Actions

Acute phase protein that is part of the coagulation cascade of proteins.
Fibrinogen is an acute phase protein that is part of the coagulation cascade of proteins. The end result of the cascade is the production of thrombin that converts fibrinogen to fibrin. Thrombin rapidly proteolyses fibrinogen, releasing fibrinopeptide A. The loss of this small peptide is not sufficient to make the resulting fibrin molecule insoluble, but it tends to form complexes with adjacent fibrin and fibrinogen molecules. Thrombin then cleaves a second peptide, fibrinopeptide B, from fibrin and the fibrin monomers formed then polymerize spontaneously to form an insoluble gel. The polymerized fibrin is held together by noncovalent and electrostatic forces and stabilized by the transamidating enzyme, factor XIIIa, that is produced by the action of thrombin on factor XIII. The insoluble fibrin aggregates (clots) and aggregated platelets then block the damaged blood vessel and prevent further bleeding. The amount of fibrinogen in the plasma can serve as a nonspecific indicator of whether or not an inflammatory process is present in the body. Fibrinogen from any mammalian source will be cleaved by thrombin from any mammalian source.

Analysis Note

Contains ~12% sodium citrate and ~18% sodium chloride.
Protein determined by biuret.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F8630341576-M341573
technique(s)

cell culture | mammalian: suitable, immunoblotting: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

biological source

rat plasma

biological source

bovine plasma

biological source

human plasma

biological source

-

concentration

60-80% (biuret)

concentration

65-85% (biuret)

concentration

-

concentration

-

mol wt

α-chain 63.5 kDa, γ chain 47 kDa (about 4% carbohydrate content), β-chain 56 kDa, soluble dimer 340 kDa

mol wt

α-chain 63.5 kDa, β-chain 56 kDa, γ chain 47 kDa (about 4% carbohydrate content), soluble dimer 340 kDa

mol wt

-

mol wt

-

form

powder

form

powder

form

lyophilized

form

lyophilized

UniProt accession no.

P02680, P14480, P06399

UniProt accession no.

P02672, P02676, P12799

UniProt accession no.

-

UniProt accession no.

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Cell Transplantation as a Method to Investigate Spinal Cord Regeneration in Regenerative and Nonregenerative Xenopus Stages
Mendez-Olivos EE and LarraIn J
Cold Spring Harbor Protocols (2018)
Optimization of trophic support for neural stem cell grafts in sites of spinal cord injury
Robinson J and Lu P
Experimental Neurology, 291, 87-97 (2017)
Salmon fibrin glue in rats: antibody studies
Laidmae I, et al.
Biologicals : Journal of the International Association of Biological Standardization, 40(1), 55-60 (2012)



Global Trade Item Number

SKUGTIN
F6755-25MG04061833261774
F6755-100MG04061833261767

Questions

Reviews

No rating value

Active Filters