Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

341576-M

Sigma-Aldrich

Fibrinogen, Human Plasma

Fibrinogen, CAS 9001-32-5, is a soluble plasma glycoprotein that is synthesized in the hepatic parenchymal cells. Contains >90% clottable protein.

Synonym(s):

Fibrinogen, Human Plasma, Coagulation Factor I

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

human plasma

Quality Level

description

Merck USA index - 14, 4071

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

sterile distilled water: soluble

storage temp.

2-8°C

General description

Do not disturb until the fibrinogen is completely solubilized.
Native fibrinogen from human plasma. A plasma glycoprotein, essential for clotting blood. Synthesized and secreted by hepatic parenchymal cells. Fibrinogen, also known as Coagulation Factor I, is an acute-phase plasma protein found at a concentration of about 300 mg/100 ml. Conversion of soluble fibrinogen to the insoluble clot-forming fibrin is the terminal stage of blood clotting. Clinically, fibrinogen is found at elevated levels during pregnancy, after surgical trauma, or in cases of myocardial infarction, and is decreased in cases of liver cirrhosis and serum hepatitis.

Biochem/physiol Actions

Contains ≥90% clottable proteins.

Warning

Toxicity: Harmful (C)

Physical form

Lyophilized from 20 mM sodium citrate-HCl, pH 7.4.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Do not store solutions on ice; precipitation will occur. Stock solutions are stable for up to 6 months at -70°C. Avoid freeze-thaw cycles.
Protein must be thawed in a 37°C water bath. Solutions used to reconstitute fibrinogen should first be brought to 37°C. Once at 37°C, the solution can be added to the fibrinogen and this mixture should remain at 37°C until the fibrinogen is completely solubilized. If the vial is disturbed before the fibrinogen is completely dissolved, the fibrinogen will precipitate. Please see vial label for lot-specific reconstitution volume.

Analysis Note

homogeneous by SDS-PAGE.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service