Skip to Content
MilliporeSigma

C4024

Carbamazepine

≥98% (HPLC), powder, sodium channel inhibitor

Synonym(s):

5H-Dibenz[b,f]azepine-5-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H12N2O
CAS Number:
Molecular Weight:
236.27
UNSPSC Code:
12352200
eCl@ss:
34051404
PubChem Substance ID:
NACRES:
NA.77
EC Number:
206-062-7
MDL number:
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Carbamazepine, powder

form

powder

Quality Segment

mp

191-192 °C (lit.)

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 29 mg/mL, H2O: insoluble, ethanol: soluble, propylene glycol: soluble

originator

Novartis

storage temp.

2-8°C

SMILES string

NC(=O)N1c2ccccc2C=Cc3ccccc13

InChI

1S/C15H12N2O/c16-15(18)17-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)17/h1-10H,(H2,16,18)

InChI key

FFGPTBGBLSHEPO-UHFFFAOYSA-N

Application

Carbamazepine has been used to investigate its behavior in wastewater treatment and during groundwater infiltration. It has also been used to find out its effects on spontaneous seizures in rats.

Biochem/physiol Actions

Carbamazepine is considered as a safe antiepileptic drug. It decreases the multiple drug therapy and maintains a better control of epilepsy.
Anticonvulsant; ligand for the GABAA receptor benzodiazepine modulatory site. Sodium channel inhibitor.

Features and Benefits

This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


Still not finding the right product?

Explore all of our products under Carbamazepine


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1A - Resp. Sens. 1 - Skin Sens. 1A - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library