Skip to Content
MilliporeSigma

Skip To

C2932

Chelerythrine chloride

plant, ≥95% (TLC), protein kinase C inhibitor, powder

Synonym(s):

1,2-Dimethoxy-N-methyl(1,3)benzodioxolo(5,6-c)phenanthridinium chloride, Toddaline chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H18ClNO4
CAS Number:
Molecular Weight:
383.82
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥95% (TLC)
Form:
powder
Quality level:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Chelerythrine chloride, ≥95% (TLC), powder

biological source

plant

Quality Level

assay

≥95% (TLC)

form

powder

color

yellow to orange

mp

213.0-214.0  °C

solubility

DMSO: 2 mg/mL

storage temp.

−20°C

SMILES string

Cl.COc1ccc2-c3ccc4cc5OCOc5cc4c3[N](C)=Cc2c1OC

InChI

1S/C21H18NO4.ClH/c1-22-10-16-13(6-7-17(23-2)21(16)24-3)14-5-4-12-8-18-19(26-11-25-18)9-15(12)20(14)22;/h4-10H,11H2,1-3H3;1H

InChI key

SUPBMPBJXZDANZ-UHFFFAOYSA-N

General description

Chelerythrine chloride (CHE), a benzophenanthridine alkaloid is an inhibitor of protein kinase C (PKC). It is the active component of Macleaya cordata.

Application

Chelerythrine chloride has been used:
  • as a supplement in heat-inactivated late-embryo extract or late-embryo extract to inhibit protein kinase C (PKC) activity
  • for in vitro Xenopus experiments
  • as a PKC inhibitor in HL-1 cells, to block the PKC pathway to study its effects on doxazosin-induced galectin-3 and collagen expression

Biochem/physiol Actions

Chelerythrine chloride (CHE) is capable of preventing the growth and apoptosis in human gastric cancer BGC-823 cells. It possesses several biological effects and also can induce the phosphorylation of a ~20 kDa protein present in the mitochondrial fraction of the rat retina.

Features and Benefits

This compound is featured on the P2 Receptors: P2X Ion Channel Family and PKC pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHL89896Y0002288220285
assay

≥95% (TLC)

assay

-

assay

-

assay

≥97% (HPLC)

form

powder

form

-

form

-

form

solid

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

100

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

solubility

DMSO: 2 mg/mL

solubility

-

solubility

-

solubility

water: 1 mg/mL, DMSO: 10 mg/mL

color

yellow to orange

color

-

color

-

color

light yellow to yellow


Still not finding the right product?

Explore all of our products under Chelerythrine chloride


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Chelerythrine chloride from Macleaya cordata induces growth inhibition and apoptosis in human gastric cancer BGC-823 cells
Zhang Z, et al.
Acta Pharmaceutica Sinica. B, 2(5), 464-471 (2012)
Protein kinase C promotes cardiac fibrosis and heart failure by modulating galectin-3 expression
Song X, et al.
Biochimica et Biophysica Acta - Molecular Cell Research, 1853(2), 513-521 (2015)
Shi-Yang Li et al.
Scientific reports, 7(1), 15317-15317 (2017-11-12)
The present study was performed to explore the role of galanin and galanin receptor 1 (GalR 1) in nociceptive modulation in the central nucleus of amygdala (CeA) in normal rats and rats with neuropathy, and the involvement of GalR 1



Global Trade Item Number

SKUGTIN
C2932-1MG04061826131985
C2932-5MG04061833484920

Questions

Reviews

No rating value

Active Filters