Skip to Content
MilliporeSigma

Skip To

220285

Chelerythrine Chloride

Naturally-occurring alkaloid.

Synonym(s):

Chelerythrine Chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H18ClNO4
CAS Number:
Molecular Weight:
383.82
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥97% (HPLC)
Form:
solid
Storage condition:
OK to freeze
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Merck USA index - 14, 2051

Quality Segment

assay

≥97% (HPLC)

form

solid

potency

660 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

light yellow to yellow

solubility

water: 1 mg/mL, DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[Cl-].[n+]1(c2c(c5c(c1)c(c(cc5)OC)OC)ccc3c2cc4c(c3)OCO4)C

InChI

1S/C21H18NO4.ClH/c1-22-10-16-13(6-7-17(23-2)21(16)24-3)14-5-4-12-8-18-19(26-11-25-18)9-15(12)20(14)22;/h4-10H,11H2,1-3H3;1H/q+1;/p-1

InChI key

WEEFNMFMNMASJY-UHFFFAOYSA-M

General description

Naturally occurring alkaloid. Cell-permeable, selective inhibitor of protein kinase C (IC50 = 660 nM). Acts on the catalytic domain irrespective of the attachment of the regulatory domain. Material is a competitive inhibitor with respect to the phosphate acceptor and a non-competitive inhibitor with respect to ATP. Over ten-fold more potent than H-7, HCl (Cat. No. 371955). Inhibits thromboxane formation and phosphoinositide metabolism in platelets. Also induces apoptotic DNA fragmentation and cell death in HL-60 human promyelocytic leukemia cells.
Naturally-occurring alkaloid. Potent, selective, and cell-permeable inhibitor of protein kinase C (IC50 = 660 nM). Acts on the catalytic domain of PKC. A competitive inhibitor with respect to the phosphate acceptor and a non-competitive inhibitor with respect to ATP. Over ten-fold more potent than H-7, HCl (Cat. No. 371955 ). Inhibits thromboxane formation and phosphoinositide metabolism in platelets. Also induces apoptotic DNA fragmentation and cell death in HL-60 human promyelocytic leukemia cells.

Biochem/physiol Actions

Product does not compete with ATP.
Reversible: no
Primary Target
PKC
Cell permeable: yes

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.
Heating to 40-50°C with gentle agitation may be necessary to achieve complete solubilization. Further dilute with buffer just prior to use.

Other Notes

Kandasamy, R.A., et al. 1995. J. Biol. Chem.270 29209.
Jarvis, W.D., et al. 1994. Cancer Res.54, 1707.
Barg, J., et al. 1992. J. Neurochem.59, 1145.
Herbert, J.M., et al. 1990. Biochem. Biophys. Res. Commun.172, 993.
Ko, F., et al. 1990. Biochim. Biophys. Acta1052, 360.
Walterova, D., et al. J. Med. Chem.24, 1100.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Harmful (C)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C2932300260PHL89896
form

solid

form

powder

form

crystalline solid

form

-

assay

≥97% (HPLC)

assay

≥95% (TLC)

assay

≥97% (HPLC)

assay

-

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

Calbiochem®

manufacturer/tradename

PhytoLab

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage condition

OK to freeze

storage condition

-

storage condition

OK to freeze, protect from light

storage condition

-


Still not finding the right product?

Explore all of our products under Chelerythrine Chloride


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
220285-5MG07790788048761

Questions

Reviews

No rating value

Active Filters