Skip to Content
MilliporeSigma

Skip To

79911

Kollidon® SR

Synonym(s):

Polyvinyl acetate-Polyvinylpyrrolidone mixture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.25
UNSPSC Code:
12162002
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

composition

polyvinylacetate, 76.0-82.0% (saponification value * 0.1534), povidone, 18.0-21.0 (N content/0.126)

impurities

≤100 ppm vinyl acetate, ≤5000 ppm residual solvents (acetic acid, acetate, formic acid (GLC)

ign. residue

≤1.7% (verified on random samples only)

loss

≤4.0% loss on drying

pH

4.0-5.0 (10% in H2O)

General description

The Kollidon products consist of soluble and insoluble grades of polyvinylpyrrolidone (PVP) of various molecular weights and particle sizes. The polymers are used as dry binders, film-formers, stabilizers in suspensions, dispersants for pigments, enzyme stabilizers, and for improvement of bioavailability.

Application

Kollidon SR is a blend of polyvinyl acetate and povidone (K 30) in the ratio 8:2 used in the pharmaceutical industry as a matrix-forming agent. This research grade product is intended for use in R&D and development only.

Legal Information

Kollidon is a registered trademark of BASF SE

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
0228696329PVP10
form

powder

form

-

form

dispersion

form

powder

ign. residue

≤1.7% (verified on random samples only)

ign. residue

≤0.1% (verified on random samples only)

ign. residue

-

ign. residue

-

pH

4.0-5.0 (10% in H2O)

pH

3.0-5.0

pH

3.0-5.5

pH

-

impurities

≤100 ppm vinyl acetate, ≤5000 ppm residual solvents (acetic acid, acetate, formic acid (GLC)

impurities

≤1 ppm hydrazine, ≤10 ppm heavy metals (verified on random samples only), ≤10 ppm lead (verified on random samples only), ≤10 ppm vinylpyrrolidinone, ≤3.0% 2-pyrrolidone, ≤400 ppm peroxides (as H2O2), ≤5.0% water, ≤500 ppm aldehydes (as acetaldehyde), ≤5000 ppm formic acid, 12.0-12.8% nitrogen (anhydrous basis)

impurities

≤0.5% sulfated ash (verified on random samples only), ≤0.500% particulate matter, agglomerates, ≤100 ppm residual monomer vinyl acetate, ≤15000 ppm acetic acid, ≤20 ppm heavy metals (verified on random samples only), ≤4.0% povidone (N content/0.126)

impurities

-

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

200

composition

polyvinylacetate, 76.0-82.0% (saponification value * 0.1534), povidone, 18.0-21.0 (N content/0.126)

composition

-

composition

-

composition

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters