Skip to Content
MilliporeSigma

02286

Kollidon® 25

Synonym(s):

Polyvinylpyrrolidone, Polyvinylpyrrolidone K25, PVP, Polyvidone, Povidone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C6H9NO)n
CAS Number:
NACRES:
NA.25
UNSPSC Code:
12162002
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

impurities

≤1 ppm hydrazine, ≤10 ppm heavy metals (verified on random samples only), ≤10 ppm lead (verified on random samples only), ≤10 ppm vinylpyrrolidinone, ≤3.0% 2-pyrrolidone, ≤400 ppm peroxides (as H2O2), ≤5.0% water, ≤500 ppm aldehydes (as acetaldehyde), ≤5000 ppm formic acid, 12.0-12.8% nitrogen (anhydrous basis)

ign. residue

≤0.1% (verified on random samples only)

pH

3.0-5.0

suitability

complies for Infrared spectrum

application(s)

sample preservation

SMILES string

C=CN1CCCC1=O

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

Application

The Kollidon products consist of soluble and insoluble grades of polyvinylpyrrolidone (PVP) of various molecular weights and particle sizes. It is used as dry binders, film-formers, stabilizers in suspensions, dispersants for pigments, enzyme stabilisiers and bioavailablility improvers.
Kollidon SR is a blend of polyvinyl acetate and povidone (K 30) in the ratio 8:2 for use as matrix-forming agent. This research grade product is intended for use in R&D and development only.

Other Notes

Polyvinylpyrrolidone is a component of Denhardt′s Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.

Legal Information

Kollidon is a registered trademark of BASF SE


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library