Skip to Content
MilliporeSigma

Skip To

STS0202

ECO TWEEN® 60

greener alternative

non-ionic detergent

Synonym(s):

TWEEN® 60, Polyethylene glycol sorbitan monostearate, Polyoxyethylene sorbitan monostearate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.28
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

mol wt

estimated mol wt 1309

composition

stearic acid, 40-60% , total stearic and palmitic acid, ≥90%

greener alternative product characteristics

Use of Renewable Feedstocks
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

CMC

27 mg/L

density

1.044 g/mL at 25 °C (lit.)

HLB

14.9

greener alternative category

SMILES string

O1C(C(C(C1)OCCO)OCCO)C(OCCO)COCCOC(=O)CCCCCCCCCCC

InChI

1S/C26H50O10/c1-2-3-4-5-6-7-8-9-10-11-24(30)34-19-18-31-20-22(32-15-12-27)26-25(35-17-14-29)23(21-36-26)33-16-13-28/h22-23,25-29H,2-21H2,1H3

InChI key

HMFKFHLTUCJZJO-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biobased surfactant and is aligned with the 7th principle of Green Chemistry "Use of Renewable Feedstocks" and the 10th principle "Design for Degradation".

TWEEN 60 is a niosome.

Application

Polyethylene glycol sorbitan monostearate, or TWEEN 60, has been used in a study to assess encapsulation of functional lipophilic components in surfactant-based colloidal delivery systems. [1] It has also been used in a study to investigate doxorubicin loaded magneto-niosomes for targeted drug delivery. [2]

Features and Benefits

  • 100 % Renewable
  • 100 % Bio-based
  • Certified to the USDA BioPreferred Program
  • Lower carbon footprint than petrochemical-based versions

Legal Information

TWEEN is a registered trademark of Croda International PLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P16298.221868.17076
greener alternative category

, Aligned

greener alternative category

-

greener alternative category

-

greener alternative category

-

composition

stearic acid, 40-60% , total stearic and palmitic acid, ≥90%

composition

stearic acid, 40-60% , total stearic and palmitic acid, ≥90%

composition

-

composition

-

mol wt

estimated mol wt 1309

mol wt

estimated mol wt 1309

mol wt

-

mol wt

-

sustainability

Greener Alternative Product

sustainability

-

sustainability

-

sustainability

-

HLB

14.9

HLB

14.9

HLB

-

HLB

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

500


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Khalid Ziani et al.
Food chemistry, 134(2), 1106-1112 (2012-10-31)
The fabrication and stability of surfactant-based colloidal delivery systems (microemulsions and emulsions) suitable for encapsulation of lipophilic active agents (vitamins and flavours) was investigated. An emulsion titration method was used to study the influence of surfactant type (Tween 20, 60
Lorena Tavano et al.
Colloids and surfaces. B, Biointerfaces, 102, 803-807 (2012-10-31)
In chemotherapy the magnetic drug targeting to a specific organ or tissue is proposed on the assumption that magnetic fields are harmless to biological systems. In this light we have vehiculated doxorubicin as model drug by novel magneto-niosomes in order
Physicochemical properties and skin permeation of [TM="Span"] 60/Tween 60 niosomes of ellagic acid
Burapacheep, V., et al.
International Journal of Pharmaceutics, 432, 9-9 (2012)



Global Trade Item Number

SKUGTIN
STS0202-1L04065266006667
STS0202-500ML04065266006674

Questions

Reviews

No rating value

Active Filters