Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.22186

Sigma-Aldrich

TWEEN® 60

for synthesis

Synonym(s):

Polyoxyethylene sorbitan monostearate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352104
EC Index Number:
500-020-4
NACRES:
NA.22

vapor pressure

<10 hPa ( 20 °C)

Quality Level

form

gel form

potency

>38000 mg/kg LD50, oral (Rat)

pH

5.5-7.7 (25 °C, 50 g/L in H2O)

kinematic viscosity

170-320 cSt(40 °C)

transition temp

flash point >149 °C

density

1.1 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O1C(C(C(C1)OCCO)OCCO)C(OCCO)COCCOC(=O)CCCCCCCCCCC

InChI

1S/C26H50O10/c1-2-3-4-5-6-7-8-9-10-11-24(30)34-19-18-31-20-22(32-15-12-27)26-25(35-17-14-29)23(21-36-26)33-16-13-28/h22-23,25-29H,2-21H2,1H3

InChI key

HMFKFHLTUCJZJO-UHFFFAOYSA-N

Application

Tween® 60 for synthesis. CAS 9005-67-8, pH 5.5 - 7.7 (50 g/l, H₂O, 25 °C).

Analysis Note

Hydroxyl value: 81 - 96
Saponification value: 45 - 55
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service