Skip to Content
MilliporeSigma

Skip To

69799

(−)-Ethyl L-lactate

greener alternative

purum, ≥98.0% (sum of enantiomers, GC)

Synonym(s):

(−)-Ethyl (S)-2-hydroxypropionate, (S)-(−)-2-Hydroxypropionic acid ethyl ester, L-Lactic acid ethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C5H10O3
CAS Number:
Molecular Weight:
118.13
UNSPSC Code:
12352108
NACRES:
NA.22
PubChem Substance ID:
EC Number:
211-694-1
Beilstein/REAXYS Number:
1720839
MDL number:
Assay:
≥98.0% (sum of enantiomers, GC)
Form:
liquid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

4.1 (vs air)

Quality Segment

vapor pressure

2 mmHg ( 20 °C), 5 mmHg ( 30 °C)

grade

purum

assay

≥98.0% (sum of enantiomers, GC)

form

liquid

optical purity

enantiomeric excess: ≥97.5:2.5

autoignition temp.

752 °F

expl. lim.

1.5 %, 100 °F

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.4130 (lit.), n20/D 1.413

bp

154 °C (lit.)

mp

−26 °C (lit.)

density

1.034 g/mL at 20 °C (lit.)

functional group

ester, hydroxyl

greener alternative category

SMILES string

C[C@H](O)C(OCC)=O

InChI

1S/C5H10O3/c1-3-8-5(7)4(2)6/h4,6H,3H2,1-2H3/t4-/m0/s1

InChI key

LZCLXQDLBQLTDK-BYPYZUCNSA-N

Features and Benefits

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is a safer and more sustainable alternative to ethyl acetate and acetone. It is an ester of natural L-lactic acid, which is produced by fermentation of sugar and thus aligns with "Safer Solvents and Auxiliaries" and "Use of Renewable Feedstocks". Click here for more information.

Other Notes

Important starting material for the synthesis of many chiral building blocks, e.g. (S)-1,2-propanediol and (S)-propylene oxide; O-Protection and reduction to the aldehyde

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
77367E3410208996
assay

≥98.0% (sum of enantiomers, GC)

assay

≥99.0% (Sum of enantiomers, GC)

assay

98%

assay

≥98.5% (GC)

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

mp

−26 °C (lit.)

mp

−26 °C (lit.)

mp

−26 °C (lit.)

mp

−26 °C (lit.)

refractive index

n20/D 1.4130 (lit.)

refractive index

n20/D 1.4130 (lit.)

refractive index

n20/D 1.4130 (lit.)

refractive index

n20/D 1.410-1.420, n20/D 1.4130 (lit.)

density

1.034 g/mL at 20 °C (lit.)

density

1.034 g/mL at 20 °C (lit.)

density

1.034 g/mL at 20 °C (lit.), 1.042 g/mL at 25 °C

density

1.034 g/mL at 20 °C (lit.)

bp

154 °C (lit.)

bp

154 °C (lit.)

bp

154 °C (lit.)

bp

154 °C (lit.)


Still not finding the right product?

Explore all of our products under (−)-Ethyl L-lactate


signalword

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

128.8 °F - closed cup

flash_point_c

53.8 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



S.D. Burke et al.
Tetrahedron Letters, 28, 4143-4143 (1987)
L. Banfi et al.
The Journal of Organic Chemistry, 49, 3784-3784 (1984)
J. Gombos et al.
Chemische Berichte, 109, 2645-2645 (1976)



Global Trade Item Number

SKUGTIN
69799-250ML04061832793986
69799-1L04061837642081

Questions

Reviews

No rating value

Active Filters