Skip to Content
MilliporeSigma
All Photos(1)

Documents

08996

Supelco

(−)-Ethyl L-lactate

analytical standard

Synonym(s):

(−)-Ethyl (S)-2-hydroxypropionate, (S)-(−)-2-Hydroxypropionic acid ethyl ester, L-Lactic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O3
CAS Number:
Molecular Weight:
118.13
Beilstein:
1720839
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.1 (vs air)

vapor pressure

2 mmHg ( 20 °C)
5 mmHg ( 30 °C)

Assay

≥98.5% (GC)

optical activity

[α]/D 10.5±1° (neat)

autoignition temp.

752 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

1.5 %, 100 °F

refractive index

n20/D 1.410-1.420
n20/D 1.4130 (lit.)

bp

154 °C (lit.)

mp

−26 °C (lit.)

density

1.034 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

C[C@H](O)C(OCC)=O

InChI

1S/C5H10O3/c1-3-8-5(7)4(2)6/h4,6H,3H2,1-2H3/t4-/m0/s1

InChI key

LZCLXQDLBQLTDK-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

128.8 °F - closed cup

Flash Point(C)

53.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuzhen Guo et al.
Lab on a chip, 15(7), 1759-1764 (2015-02-17)
A simple method to fabricate microchannels is demonstrated based on an inkjet printing liquid template. The morphology of the liquid template can be well controlled by using ink with viscosity sensitive to temperature. The as-prepared Y-shape microchannel is used as
Stéphanie Rollero et al.
Applied microbiology and biotechnology, 99(5), 2291-2304 (2014-11-22)
Volatile compounds produced by yeast during fermentation greatly influence the organoleptic qualities of wine. We developed a model to predict the combined effects of initial nitrogen and phytosterol content and fermentation temperature on the production of volatile compounds. We used
Zuobing Xiao et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 978-979, 122-130 (2014-12-30)
A simple, rapid and solvent-free multi-residue method has been developed and applied to confirm and quantify a series of volatile compounds in five cherry wines by gas chromatography coupled with mass spectrometry (GC-MS). Four parameters (e.g., coating material of fiber
Pingxi Xu et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(46), 16592-16597 (2014-10-29)
Insect repellents are important prophylactic tools for travelers and populations living in endemic areas of malaria, dengue, encephalitis, and other vector-borne diseases. DEET (N,N-diethyl-3-methylbenzamide) is a 6-decade-old synthetic repellent, which is still considered the gold standard of mosquito repellents. Mosquitoes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service