Skip to Content
MilliporeSigma

Skip To

36932

Hexadecyltrimethylammonium bromide

analytical standard

Synonym(s):

CTAB, Cetrimonium bromide, Cetyltrimethylammonium bromide, Palmityltrimethylammonium bromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)15N(Br)(CH3)3
CAS Number:
Molecular Weight:
364.45
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
200-311-3
Beilstein/REAXYS Number:
3598189
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

description

cationic

assay

98-102%

shelf life

limited shelf life, expiry date on the label

mol wt

364.45 g/mol

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

mp

248-251 °C (lit.)

solubility

H2O: soluble 36.4 g/L at 20 °C (completely)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C19H42N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1H/q+1;/p-1

InChI key

LZZYPRNAOMGNLH-UHFFFAOYSA-M

General description

Cetrimonium bromide(CTAB) is a quaternary ammonium derivative known to enhance anti-tumor activity. It is one of the components of the broad-spectrum antiseptic cetrimide and may also be used as a buffer for DNA extraction.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

CTAB may be used as a reference standard for the determination of:
  • CTAB in various water samples by combining a dispersive liquid-liquid microextraction (DLLME) technique with high performance liquid chromatography.
  • CTAB in pharmaceutical formulations by reversed-phase high-performance liquid chromatography (RP- HPLC).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
52367H588252370
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

ion pair chromatography: suitable

technique(s)

DNA extraction: suitable, RNA extraction: suitable

technique(s)

UV/Vis spectroscopy: suitable, electrophoresis: suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

application(s)

-

application(s)

genomic analysis

application(s)

detection

format

neat

format

-

format

-

format

-

Quality Level

100

Quality Level

100

Quality Level

300

Quality Level

200

grade

analytical standard

grade

-

grade

-

grade

-

assay

98-102%

assay

≥99.0% (AT)

assay

≥98%

assay

≥96.0% (AT)


Still not finding the right product?

Explore all of our products under Hexadecyltrimethylammonium bromide


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

target_organs

Gastrointestinal tract, Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

471.2 °F - closed cup

flash_point_c

244 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Potential use of cetrimonium bromide as an apoptosis-promoting anti-cancer agent for head and neck cancer
Ito E, et al.
Molecular Pharmacology (2009)
Journal of Applied Chemistry Application of dispersive liquid-liquid microextraction and high-performance liquid chromatography for the determination of cetrimonium bromide in water samples.
Rajabi M and Ghazaghi M
J. Appl. Chem., 8(27) (2013)
Development and validation of RP-HPLC method for cetrimonium bromide and lidocaine determination.
Malenovic A, et al.
Il Farmaco (Societa Chimica Italiana : 1989), 60(2), 157-161 (2005)



Global Trade Item Number

SKUGTIN
36932-1G04061831833058

Questions

Reviews

No rating value

Active Filters