Skip to Content
MilliporeSigma
Hexadecyltrimethylammonium bromide USP/NF

Hexadecyltrimethylammonium bromide

Hexadecyltrimethylammonium bromide USP/NF
Synonym(s):
CTAB, Cetrimonium bromide, Cetyltrimethylammonium bromide, Palmityltrimethylammonium bromide, Hexadecyltrimethylammonium bromide
Linear Formula:
CH3(CH2)15N(Br)(CH3)3
CAS Number:
Molecular Weight:
364.45
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Reference Material Type
Special Grade
Technique