Skip to Content
MilliporeSigma

Skip To

04067

Fmoc-6-Ahx-OH

≥97.0% (HPLC)

Synonym(s):

6-(Fmoc-amino)caproic acid, 6-(Fmoc-amino)hexanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H23NO4
CAS Number:
Molecular Weight:
353.41
UNSPSC Code:
12352209
NACRES:
NA.26
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
5883220
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥97.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

color

white

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)CCCCCNC(=O)OCC1c2ccccc2-c3ccccc13

InChI

1S/C21H23NO4/c23-20(24)12-2-1-7-13-22-21(25)26-14-19-17-10-5-3-8-15(17)16-9-4-6-11-18(16)19/h3-6,8-11,19H,1-2,7,12-14H2,(H,22,25)(H,23,24)

InChI key

FPCPONSZWYDXRD-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
040660406804069
reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

functional group

Fmoc

functional group

Fmoc

functional group

Fmoc

functional group

Fmoc

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

100

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

form

powder

form

powder

form

powder

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Sergej Karel et al.
Carbohydrate polymers, 201, 300-307 (2018-09-23)
New materials based on molecules naturally occurred in body are assumed to be fully biocompatible and biodegradable. In our study, we used hyaluronic acid (HA) modified with peptides, which meet all this criterion and could be advantageously used in tissue
Jing Han et al.
Molecular pharmaceutics, 15(7), 2840-2856 (2018-05-26)
GLP-1 analogs suffer from the main disadvantage of a short in vivo half-life. Lithocholic acid (LCA), one of the four main bile acids in the human body, possesses a high albumin binding rate. We therefore envisioned that a LCA-based peptide
Christos Kontos et al.
Nature communications, 11(1), 5981-5981 (2020-11-27)
Targeting a specific chemokine/receptor axis in atherosclerosis remains challenging. Soluble receptor-based strategies are not established for chemokine receptors due to their discontinuous architecture. Macrophage migration-inhibitory factor (MIF) is an atypical chemokine that promotes atherosclerosis through CXC-motif chemokine receptor-4 (CXCR4). However



Global Trade Item Number

SKUGTIN
130109-1KG04061838726827
185507-100G04061835554461
185507-5G04061835554478
822290025204061839612723
822290025104061839612716
185507-500G04061838757272
21-0210-5-500G-J04061832685403
822290999904022536487960
822290100004022536487953
822290025004022536487946
70450-100G04061832826431
130109-250G04061838726834
70450-500G04061833439531
70448-5G04061833358672
04067-5G04061838628800
04067-1G04061838628794

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters