Skip to Content
MilliporeSigma

04066

Fmoc-5-Ava-OH

≥98.0% (HPLC)

Synonym(s):

5-(Fmoc-amino)pentanoic acid, 5-(Fmoc-amino)valeric acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H21NO4
CAS Number:
Molecular Weight:
339.39
UNSPSC Code:
12352209
NACRES:
NA.26
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3565625
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

color

white

mp

137-145 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)CCCCNC(=O)OCC1c2ccccc2-c3ccccc13

InChI

1S/C20H21NO4/c22-19(23)11-5-6-12-21-20(24)25-13-18-16-9-3-1-7-14(16)15-8-2-4-10-17(15)18/h1-4,7-10,18H,5-6,11-13H2,(H,21,24)(H,22,23)

InChI key

ULLSWWGYZWBPHK-UHFFFAOYSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library