Skip to Content
MilliporeSigma

Skip To

SCT142

BioTracker 488 Green Microtubule Cytoskeleton Dye

Live cell imaging microtubule dye for imaging the cytoskeleton of living cells

Synonym(s):

Live cell imaging probe

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.47
UNSPSC Code:
12352207
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


technique(s)

cell based assay: suitable

detection method

fluorometric

shipped in

ambient

storage temp.

2-8°C

General description

Cytoskeleton proteins such as actin and tubulin play an important role in cell structure and motility. Dysregulation of cytoskeletal proteins are also involved in cancer metastasis and disease. Taxol binds to polymerized tubulin and stabilizes microtubules, resulting in inhibition of mitosis. However, fluorescent taxol compounds like the BioTracker Microtubule Cytoskeleton dyes are less disruptive of microtubule dynamics and cell division, presumably due to lower binding affinity of the fluorescent probe compared to taxol itself.

The BioTracker Cytoskeleton Microtubule Dye is a green fluorescent cell-permeable taxol probe for imaging the microtubule cytoskeleton in live cells. It is a simple, rapid and sensitive stain that can be detected using the FITC channel. The dye is supplied with a vial of 100mM verapamil, an efflux pump inhibitor that may improve probe retention and staining in certain cell types.

Spectral Properties
Absorbance: 500nm
Emission: 515nm

Application

Live cell fluorescent imaging
Live cell imaging microtubule dye for imaging the cytoskeleton of living cells
Research Category
Cell Signaling
Research Sub Category
Live Cell Dye

Physical form

Liquid

Preparation Note

Store BioTracker 488 Green Microtubule Cytoskeleton Dye at 2-8ºC. Protect From Light.
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.

Analysis Note

Spectral Properties
Absorbance: 500nm
Emission: 515nm

Other Notes

1) 1 vial of 50µL BioTracker 488 Green Microtubule Cytoskeleton Dye (1000X in DMSO) (CS224593)

2) 1 vial of 100µL Verapamil HCL (100mM in DMSO) (CS224592)

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCT120SCT136SCT135
technique(s)

cell based assay: suitable

technique(s)

cell based assay: suitable

technique(s)

cell based assay: suitable

technique(s)

cell based assay: suitable

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

detection method

fluorometric

detection method

fluorometric

detection method

fluorometric

detection method

fluorometric


pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class

10 - Combustible liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Questions

  1. Would it be advisable to employ any form of stimulus to induce microtubular organization in cancer cells when treating them with our drug to inhibit migration and analyze cytoskeletal changes, given that we have acquired the green microtubule dye?

    1 answer
    1. This dye can be helpful in visualizing the microtubules of a cell. If you are studying a cancer cell line, then the natural dynamics of the microtubules will occur as the cells go through their cycle and divide. It's worth considering whether you want to add an agent to induce mitosis further, especially as you are trying to see the effects of a drug on this mechanism as it occurs.

      Helpful?

Reviews

No rating value

Active Filters