Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCT135

Sigma-Aldrich

BioTracker 405 Blue Mitochondria Dye

Live cell imaging mitochondrial dye that stains the membrane of mitochondria used to detect cell viability, metabolic activity and overall cell health.

Synonym(s):

Live cell imaging probe, Live cell mitochondrial imaging probe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.47

technique(s)

cell based assay: suitable

detection method

fluorometric

shipped in

ambient

General description

Mitochondria are organelles found in large numbers in most cells, in which the biochemical processes of respiration and energy production occurs. In metabolically active cells, mitochondria produce a membrane potential by maintaining a proton gradient across the inner and outer membranes. Loss of mitochondrial membrane potential is a hallmark for apoptosis and poor cell health.

BioTracker Mitochondria dyes are fluorogenic stains for staining mitochondria in live cells. The dyes are membrane permeable and become brightly fluorescent upon accumulation in the mitochondrial membrane. Staining is lost when mitochondria become depolarized during cell death, allowing monitoring of cell viability.

The BioTracker 405 Blue Mitochondria Dye can be detected using the blue DAPI channel.

Spectral Properties
Absorbance: 398nm
Emission: 440nm

Application

Live cell fluorescent imaging
Live cell imaging mitochondrial dye that stains the membrane of mitochondria used to detect cell viability, metabolic activity and overall cell health.
Research Category
Cell Imaging
Research Sub Category
Live Cell Dye

Quality

Spectral Properties
Absorbance: 398nm
Emission: 440nm

Physical form

Lyophilized

Storage and Stability

Store BioTracker 405 Blue Mitochondria Dye at -20ºC. Protect From Light.
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service