Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NC.01
UNSPSC Code:
41105337
Pricing and availability is not currently available.
form
liquid
mol wt
10-200 kDa
packaging
pkg of 500 μL
storage temp.
−20°C
General description
The mPAGE® Unstained Protein Standard is a mixture of twelve recombinant unstained proteins ranging from 10kDa to 200kDa. The 85kDa, 25kDa, and 10kDa bands have higher intensity to serve as reference points. mPAGE® Unstained Protein Standard is suitable for molecular weight determination after SDS-PAGE or Western blotting and serves as a standard to calibrate the mobility of prestained markers.
Storage Buffer: 20 mM Tris-phosphate(pH 7.5 at 25°C), 2 % SDS, 0.2 mM Dithiothreitol, 3.6 M Urea, and 15 % (v/v) Glycerol. Load 2.5 to 10 μl per lane.
Storage Buffer: 20 mM Tris-phosphate(pH 7.5 at 25°C), 2 % SDS, 0.2 mM Dithiothreitol, 3.6 M Urea, and 15 % (v/v) Glycerol. Load 2.5 to 10 μl per lane.
- Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Application
SDS-PAGE, Western blot
- Immunoblotting (Western)
- Western Blotting
Other Notes
4 °C for up to 3 months, or 25 °C for up to two weeks.
Legal Information
MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany
1 of 1
This Item | |||
|---|---|---|---|
| form liquid | form liquid | form liquid | form solid |
| mol wt 10-200 kDa | mol wt 10-203 kDa | mol wt - | mol wt 15-600 kDa |
| packaging pkg of 500 μL | packaging pkg of 500 μL | packaging - | packaging - |
| storage temp. −20°C | storage temp. −20°C | storage temp. - | storage temp. −20°C |
Storage Class
10 - Combustible liquids
wgk
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| MPSTD3 | 04061841077640 |



