Skip to Content
MilliporeSigma

MPSTD4

mPAGE® Color Protein Standard

Synonym(s):

protein markers, protein molecular weight markers, protein standards

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

NACRES:
NC.01
UNSPSC Code:
41105337
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

liquid

mol wt

10-203 kDa

packaging

pkg of 500 μL

manufacturer/tradename

Millipore MPSTD4

storage temp.

−20°C

Application

SDS-PAGE, Western blot
  • Immunoblotting (Western)
  • Western Blotting

General description

The mPAGE® Color Protein Standard is a mixture of ten highly purified prestained proteins ranging from 10 kDa to 203 kDa, covalently coupled with different chromophores. mPAGE® Color Protein Standard is designed for observing protein separation during SDS-PAGE, verifying western transfer efficiency on membranes, and approximating the size of proteins.

Storage Buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25 °C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3, 33% glycerol. Load 2.5 to 10 μl per lane.

  • Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Other Notes

4 °C for up to 3 months, or 25 °C for up to two weeks.

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service