Skip to Content
MilliporeSigma

Skip To

8.22283

Methanol

≥99.5% (GC), for preparative purposes, for synthesis, for cleaning, for extraction, for analytical purposes, EMPLURA®

Synonym(s):

Methyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
NACRES:
NB.21
EC Index Number:
200-659-6
UNSPSC Code:
12352104
MDL number:
Beilstein/REAXYS Number:
1098229
eCl@ss:
39020101
Assay:
≥99.5% (GC)
Grade:
synthesis grade
Bp:
64.7 °C (lit.)
Vapor pressure:
128 hPa ( 20 °C), 410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Methanol, for preparative purposes, EMPLURA®

grade

synthesis grade

Quality Segment

vapor density

1.11 (vs air)

vapor pressure

128 hPa ( 20 °C), 410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)

description

for analytical purposes, for extraction

product line

EMPLURA®

assay

≥99.5% (GC)

form

liquid

autoignition temp.

725 °F

potency

5628 mg/kg LD50, oral (Rat), 17100 mg/kg LD50, skin (Rabbit)

expl. lim.

36 %

dilution

(low-cost alternative, for basic applications)

impurities

≤0.1% Water

evapn. residue

≤0.0010%

refractive index

n20/D 1.329 (lit.)

kinematic viscosity

0.54-0.59 cSt(20 °C)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

transition temp

flash point 9.7 °C

density

0.791 g/mL at 25 °C (lit.)

application(s)

environmental
food and beverages
industrial qc
pharmaceutical

format

neat

storage temp.

2-30°C

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Analysis Note

Evaporation Number: 1.9
Purity (GC): ≥ 99.5 %
Identity (IR): conforms
Density (d 20 °C/ 4 °C): 0.791 - 0.792
Evaporation residue: ≤ 0.0010 %
Water: ≤ 0.10 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.070181.945161.06009
assay

≥99.5% (GC)

assay

≥99.8% (GC)

assay

-

assay

≥99.9% (GC)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

300

application(s)

environmental
food and beverages
industrial qc
pharmaceutical

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

application(s)

pharmaceutical

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

grade

synthesis grade

grade

ACS reagent, reagent grade

grade

synthesis grade

grade

ACS reagent

form

liquid

form

liquid

form

liquid

form

liquid

refractive index

n20/D 1.329 (lit.)

refractive index

n20/D 1.329 (lit.)

refractive index

n20/D 1.329 (lit.)

refractive index

n20/D 1.329 (lit.)


Still not finding the right product?

Explore all of our products under Methanol


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
822283918004022536944234
822283902504022536944227
822283500004022536944210
822283901104027269350345
822283100004022536487649
822283250004022536487656

Questions

Reviews

No rating value

Active Filters