Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

1.07018

Supelco

Methanol

≥99.8% (GC), EMPARTA® ACS reagent, for extraction, for analytical purposes

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-659-6
NACRES:
NB.21
grade:
ACS reagent
for extraction
reagent grade
application(s):
environmental
food and beverages
industrial qc
pharmaceutical
sample preparation
bp:
64.7 °C (lit.)
vapor pressure:
128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

Product Name

Methanol, for analysis EMPARTA® ACS, for extraction

grade

ACS reagent
for extraction
reagent grade

Quality Level

vapor density

1.11 (vs air)

vapor pressure

128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

description

for analytical purposes

product line

EMPARTA®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

725 °F

potency

5628 mg/kg LD50, oral (Rat)
17100 mg/kg LD50, skin (Rabbit)

expl. lim.

36 %

dilution

(for routine analytical applications)

impurities

≤0.0002 meq/g Titrable base
≤0.0003% Substances reducing potassium permanganate (as O)
≤0.0003 meq/g Titrable acid
≤0.001% Acetaldehyde
≤0.001% Acetone
≤0.001% Formaldehyde
≤0.1% Water

evapn. residue

≤0.001%

color

clearAPHA: ≤10

refractive index

n20/D 1.329 (lit.)

kinematic viscosity

0.54-0.59 cSt(20 °C)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

transition temp

flash point 9.7 °C

density

0.791 g/mL at 25 °C (lit.)

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

format

neat

storage temp.

2-30°C

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We offer EMPARTA grade solvents for routine tasks in analytical laboratories. EMPARTA grade solvents offer fewer test parameters than EMSURE, yet are still of high quality. EMPARTA solvents meet ACS requirements, making them ideal for a wide range of analytical applications.

Application



  • A technique based on infrared spectroscopy for determining sulfanilamide levels sustainably: Progress and comparisons of greenness and whiteness using ComplexGAPI, AGREE, and RGB.: This research introduces a sustainable method using methanol in infrared spectroscopy to measure sulfanilamide levels, highlighting advancements in green analytical chemistry (Lahkar et al., 2024).


  • A novel method for quantitative determination of multiple substances using Raman spectroscopy combined with CWT.: The study utilizes methanol as a solvent in a new Raman spectroscopy technique to quantitatively analyze various substances, demonstrating enhanced precision in chemical analysis (Yang et al., 2024).


Packaging

1.07018.2521: 2.5L, Minimum order quantity: 4

Analysis Note

Purity (GC): ≥ 99.8 %
Identity (IR): conforms
Appearance: clear
Color: ≤ 10 Hazen
Solubility in water: conforms
Titrable acid: ≤ 0.0003 meq/g
Titrable base: ≤ 0.0002 meq/g
Boiling point: 64 - 65 °C
Acetone: ≤ 0.001 %
Acetaldehyde: ≤ 0.001 %
Formaldehyde: ≤ 0.001 %
Readily carbonizable substances: conforms
Substances reducing potassium permanganate (as O): ≤ 0.0003 %
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.1 %

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service