Skip to Content
MilliporeSigma

Skip To

8.20871

Sodium ethoxide

for synthesis

Synonym(s):

Sodium ethoxide, Sodium ethylate, Sodium ethanolate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
Molecular Weight:
68.05
UNSPSC Code:
12352302
EC Index Number:
205-487-5
NACRES:
NA.22
MDL number:
Form:
powder
Grade:
synthesis grade
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Segment

form

powder

autoignition temp.

30-50 °C

potency

598 mg/kg LD50, oral (Rat)

pH

13 (20 °C, 5 g/L in H2O)

density

0.87 g/cm3 at 20 °C

bulk density

300 kg/m3

SMILES string

[Na+].[O-]CC

InChI

1S/C2H5O.Na/c1-2-3;/h2H2,1H3;/q-1;+1

InChI key

QDRKDTQENPPHOJ-UHFFFAOYSA-N

General description

for synthesis

Application

Sodium ethoxide for synthesis. CAS No. 141-52-6, EC Number 205-487-5.

Analysis Note

Assay (acidimetric)≥ 95.0 %Free NaOH≤ 2.0 %

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.1819971210164992
grade

synthesis grade

grade

synthesis grade

grade

-

grade

reagent grade

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

form

powder

form

liquid

form

powder

form

powder

pH

13 (20 °C, 5 g/L in H2O)

pH

11 ( in H2O)

pH

-

pH

-

density

0.87 g/cm3 at 20 °C

density

0.88 g/cm3 at 20 °C

density

-

density

-

autoignition temp.

30-50 °C

autoignition temp.

425 °C

autoignition temp.

-

autoignition temp.

851 °F


Still not finding the right product?

Explore all of our products under Sodium ethoxide


pictograms

FlameCorrosion

signalword

Danger

Hazard Classifications

Flam. Sol. 1 - Self-heat. 1 - Skin Corr. 1A

supp_hazards

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk

WGK 1

flash_point_f

86.0 °F - closed cup

flash_point_c

30 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters