Skip to Content
MilliporeSigma
All Photos(3)

Documents

164992

Sigma-Aldrich

Sodium methoxide

reagent grade, 95%, powder

Synonym(s):

Sodium methylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOCH3
CAS Number:
Molecular Weight:
54.02
Beilstein:
3592982
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

vapor density

1.1 (vs air)

vapor pressure

50 mmHg ( 20 °C)

Assay

95%

form

powder

autoignition temp.

851 °F

expl. lim.

36 %

SMILES string

[Na+].C[O-]

InChI

1S/CH3O.Na/c1-2;/h1H3;/q-1;+1

InChI key

WQDUMFSSJAZKTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium methoxide is a sodium alkoxide. Its reaction with unactivated aryl bromides in the presence of copper catalyst under various reaction conditions has been studied. It reacts with aryl halides in the presence of a palladium catalyst in DMF to afford arenes.

Application

Sodium methoxide has been used in the preparation of 5-(3-azidopropyl) uridine. It may be used in the synthesis of methyl 2-cyano-lup-3-hydroxy-2-en-28-oate (CN-BA), a 2-cyano derivative of betulinic acid (BA).
Sodium methoxide may be used as an alkali catalyst for the production of fatty acid methyl esters (biodiesel) from vegetable oils via transesterification.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Sol. 1 - Met. Corr. 1 - Self-heat. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production of biodiesel through optimized alkaline-catalyzed transesterification of rapeseed oil.
Rashid U and Anwar F
Fuel: The Science and Technology of Fuel and Energy, 87(3), 265-273 (2008)
A review on biodiesel production using catalyzed transesterification
Leung DYC, et al
Applied Energy, 87(4), 1083-1095 (2010)
Palladium hydrides in organic synthesis. Reduction of aryl halides by sodium methoxide catalyzed by tetrakis (triphenylphosphine) palladium.
Zask A and Helquist P.
The Journal of Organic Chemistry, 43(8), 1619-1620 (1978)
The copper catalysed reaction of sodium methoxide with aryl bromides. A mechanistic study leading to a facile synthesis of anisole derivatives.
Aalten HL, et al.
Tetrahedron, 45(17), 5565-5578 (1989)
Sudhakar Chintharlapalli et al.
Carcinogenesis, 28(11), 2337-2346 (2007-08-29)
Betulinic acid (BA) is a phytochemical triterpenoid acid from bark extracts and is cytotoxic to cancer cells and tumors. We modified the A-ring of BA to give a 2-cyano-1-en-3-one moiety and the effects of the 2-cyano-lup-1-en-3-oxo-20-oic acid (CN-BA), 2-cyano derivative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service