If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf
Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product Name
N-Bromosuccinimide, for synthesis
vapor pressure
14.8 hPa ( 20 °C)
Quality Segment
form
powder
potency
>2000 mg/kg LD50, oral (Rat)
reaction suitability
reagent type: oxidant
mp
174-179 °C
solubility
14.8 g/L (decomposition)
density
2.1 g/cm3 at 20 °C
bulk density
750 kg/m3
storage temp.
2-8°C
SMILES string
BrN1C(=O)CCC1=O
InChI
1S/C4H4BrNO2/c5-6-3(7)1-2-4(6)8/h1-2H2
InChI key
PCLIMKBDDGJMGD-UHFFFAOYSA-N
Application
- Mechanistic study on iodine-catalyzed aromatic bromination of aryl ethers by N-bromosuccinimide: This study explores the roles of NBS in organic synthesis, particularly in radical and electrophilic substitutions (Pramanick et al., 2017).
- One‐Pot Protocol for the Synthesis of Imidazoles and Quinoxalines using N‐Bromosuccinimide: This paper reports a green, efficient synthesis of heterocycles mediated by NBS (Pardeshi et al., 2017).
- N-Bromosuccinimide as an Interfacial Alleviator for Br/I Exchange in Perovskite for Solar Cell Fabrication: This research demonstrates the use of NBS in improving the crystallization and defect passivation in perovskite solar cells (Pegu et al., 2021).
- N‐Bromosuccinimide‐Mediated Radical Cyclization of 3‐Arylallyl Azides: Synthesis of 3‐Substituted Quinolines: Discusses the application of NBS under visible light irradiation to synthesize quinolines (Wang et al., 2015).
- N-Bromosuccinimide mediated synthesis of triazatruxenes from indoles: This study highlights the cyclotrimerization of indoles using NBS, demonstrating its utility as a reagent in complex organic transformations (Toworakajohnkun et al., 2017).
Analysis Note
Identity (IR): passes test
1 of 1
This Item | |||
|---|---|---|---|
| form powder | form powder | form powder | form powder |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| Quality Level 200 | Quality Level 400 | Quality Level 200 | Quality Level 200 |
| solubility 14.8 g/L (decomposition) | solubility - | solubility - | solubility - |
| mp 174-179 °C | mp 146-151 °C | mp 146-151 °C | mp 175-180 °C (dec.) (lit.) |
| bulk density 750 kg/m3 | bulk density 600 kg/m3 | bulk density 600 kg/m3 | bulk density - |
Still not finding the right product?
Explore all of our products under N-Bromosuccinimide
signalword
Warning
Hazard Classifications
Aquatic Acute 1 - Eye Irrit. 2 - Met. Corr. 1 - Muta. 2 - Ox. Sol. 3 - Skin Irrit. 2 - Skin Sens. 1B
Storage Class
5.1B - Oxidizing hazardous materials
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 8019490005 | 04022536371153 |
| 8019490250 | 04022536371160 |
| 8019491000 | 04022536371177 |
| 8019499025 | 04022536371184 |
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-








