Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.45027

Sigma-Aldrich

N-Chlorosuccinimide

Msynthplus

Synonym(s):

N-Chlorosuccinimide, NCS, Succinchlorimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4ClNO2
CAS Number:
Molecular Weight:
133.53
MDL number:
UNSPSC Code:
12352111
EC Index Number:
204-878-8

Quality Level

form

powder

potency

1000 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: oxidant

bp

216.5 °C

mp

146-151 °C

transition temp

flash point >110 °C

density

1.65 g/cm3 at 20 °C

bulk density

600 kg/m3

storage temp.

2-8°C

InChI

1S/C4H4ClNO2/c5-6-3(7)1-2-4(6)8/h1-2H2

InChI key

JRNVZBWKYDBUCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Color (visual): white
Appearance of substance (visual): powder
Assay (iodometric): ≥ 98.0 %
Melting range (lower value): ≥ 146 °C
Melting range (upper value): ≤ 151 °C
Identity (IR): passes test
The analysis of residual solvents is not possible due to the physicochemical properties of the material.

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service