Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
grade
certified reference material
Quality Segment
product line
Certipur®
pH
9.0 (25 °C in H2O)
bp
100 °C/1013 hPa
density
1.00 g/cm3 at 20 °C
application(s)
environmental
food and beverages
general analytical
industrial qc
pharmaceutical
format
mixture
storage temp.
15-25°C
General description
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.
Application
Features and Benefits
- Measured and qualified at an ISO/IEC 17025 and ISO 17034 accredited laboratory
- Traceable to secondary reference standard from NIST and primary certified reference standard from PTB
- Ready-to-use solution available in 30 mL sachets
- Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an effective audit
- Product label follows the ISO Guide 31 and contains information about ordering number, article description, lot number, safety data, and expiry date
- Packaging material subjected to comprehensive stability studies as per ISO Guide 35
- Accurate up to +/- 0.03 at 25°C
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| format mixture | format mixture | format mixture | format mixture |
| product line Certipur® | product line Certipur® | product line Certipur® | product line Certipur® |
| bp 100 °C/1013 hPa | bp - | bp 109 °C/1013 hPa | bp 100 °C/1013 hPa |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| application(s) environmental | application(s) environmental | application(s) environmental | application(s) environmental |
| storage temp. 15-25°C | storage temp. 15-25°C | storage temp. 15-25°C | storage temp. 15-25°C |
Still not finding the right product?
Explore all of our products under Buffer solution
signalword
Danger
hcodes
Hazard Classifications
Repr. 1B
Storage Class
6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1990030001 | 04022536731483 |




