Skip to Content
MilliporeSigma

Skip To

1.09461

Buffer solution

(boric acid/potassium chloride/sodium hydroxide), traceable to SRM from NIST and PTB pH 9.00 (20°C) Certipur®

Synonym(s):

Buffer solution pH 9.00 (20°C)(boric acid/potassium chloride/sodium hydroxide)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.25
UNSPSC Code:
12161705
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Level

product line

Certipur®

pH

9.0 (20 °C in H2O)

density

1.0 g/cm3 at 20 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

The boric acid/potassium chloride/sodium hydroxide certified buffer solution is gravimetrically prepared using high-quality reagents as per DIN EN ISO/IEC 17025. Its pH value of 9.00 at 20°C is measured following a 5-point calibration method, using a combined glass electrode. The measurement is made following DIN 19268 using reference buffer solutions compliant to DIN 19266, NIST, IUPAC, Ph. Eur, and USP. Its pH value is strongly dependent on the temperature and therefore a constant temperature must be maintained during its use.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

The buffer solution is meant to be used as a calibration standard for pH measuring instruments or pH electrodes and as a control for measuring the pH of solutions.

Features and Benefits

  • Measured and qualified at an ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Traceable to secondary reference standards from NIST and primary certified reference standards from PTB
  • Ready-to-use solution
  • Available in a sealed plastic bottle and sustainable Titripac® packaging for a reduced waste generation
  • Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an effective audit
  • Product label follows the ISO Guide 31 and contains information about ordering number, article description, lot number, safety data, and expiry date
  • Packaging material subjected to comprehensive stability studies as per ISO Guide 35

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Titripac is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.990031.094621.99038
format

mixture

format

mixture

format

mixture

format

mixture

product line

Certipur®

product line

Certipur®

product line

Certipur®

product line

Certipur®

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

pH

9.0 (20 °C in H2O)

pH

9.0 (25 °C in H2O)

pH

11.0 (25 °C in H2O)

pH

8.0 (25 °C in H2O)


Still not finding the right product?

Explore all of our products under Buffer solution


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
109461400004027269029852
109461100004022536114644
109461901004022536114668

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters