Skip to Content
Merck

Skip To

1705323

USP

Ubidecarenone Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Coenzyme Q9, Q-9, Ubiquinone-45, Ubiquinone-9

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C54H82O4
CAS Number:
Molecular Weight:
795.23
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1900081
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

ubidecarenone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

COC1=C(OC)C(=O)C(C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(\C)C)=C(C)C1=O

InChI

1S/C54H82O4/c1-40(2)22-14-23-41(3)24-15-25-42(4)26-16-27-43(5)28-17-29-44(6)30-18-31-45(7)32-19-33-46(8)34-20-35-47(9)36-21-37-48(10)38-39-50-49(11)51(55)53(57-12)54(58-13)52(50)56/h22,24,26,28,30,32,34,36,38H,14-21,23,25,27,29,31,33,35,37,39H2,1-13H3/b41-24+,42-26+,43-28+,44-30+,45-32+,46-34+,47-36+,48-38+

InChI key

UUGXJSBPSRROMU-WJNLUYJISA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ubidecarenone Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Ubidecarenone
  • Ubidecarenone Capsules
  • Ubidecarenone Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
170530117053121011029
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

API family

ubidecarenone

API family

ubidecarenone

API family

ubidecarenone

API family

acitretin

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Leilei Li et al.
International journal of systematic and evolutionary microbiology, 64(Pt 7), 2407-2415 (2014-04-26)
Five acetic acid bacteria isolates, awK9_3, awK9_4 ( = LMG 27543), awK9_5 ( = LMG 28092), awK9_6 and awK9_9, obtained during a study of micro-organisms present in traditionally produced kefir, were grouped on the basis of their MALDI-TOF MS profile with LMG
Asif Hameed et al.
International journal of systematic and evolutionary microbiology, 64(Pt 7), 2330-2337 (2014-04-20)
Two Gram-staining-negative, aerobic, rod-shaped, non-spore-forming bacterial strains that are motile by a monopolar flagellum, designated CC-AMH-11(T) and CC-AMHZ-5, were isolated from droppings of a seashore bird off the coast of Hualien, Taiwan. The strains showed 99.7% mutual pairwise 16S rRNA
Wen-Bo Tan et al.
International journal of systematic and evolutionary microbiology, 65(Pt 1), 225-229 (2014-10-19)
A Gram-staining-negative, rod-shaped, motile and facultatively anaerobic bacterial strain, designated X2(T), was isolated from the sludge of an anaerobic, denitrifying, sulfide-removal bioreactor, and found to oxidize sulfide anaerobically with nitrate as electron acceptor. The strain grew at salinities of 0-3%



Global Trade Item Number

SKUGTIN
1705323-15MG04061833861608

Questions

Reviews

No rating value

Active Filters