Skip to Content
Merck
All Photos(1)

Key Documents

1705312

USP

Ubidecarenone for System Suitability

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Coenzyme Q10, Q-10, Ubiquinone 50, Ubiquinone-10

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H90O4
CAS Number:
Molecular Weight:
863.34
Beilstein:
1900141
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ubidecarenone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

O=C(C(OC)=C1OC)C(C/C=C(C)/CC/C=C(CC/C=C(CC/C=C(CC/C=C(CC/C=C(CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CCC=C(C)C)\C)\C)\C)\C)\C)=C(C)C1=O

InChI

1S/C59H90O4/c1-44(2)24-15-25-45(3)26-16-27-46(4)28-17-29-47(5)30-18-31-48(6)32-19-33-49(7)34-20-35-50(8)36-21-37-51(9)38-22-39-52(10)40-23-41-53(11)42-43-55-54(12)56(60)58(62-13)59(63-14)57(55)61/h24,26,28,30,32,34,36,38,40,42H,15-23,25,27,29,31,33,35,37,39,41,43H2,1-14H3/b45-26+,46-28+,47-30+,48-32+,49-34+,50-36+,51-38+,52-40+,53-42+

InChI key

ACTIUHUUMQJHFO-UPTCCGCDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ubidecarenone for System Suitability USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monograph such as Ubidecarenone

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shirin Kalyan et al.
The Journal of clinical endocrinology and metabolism, 99(4), 1307-1313 (2014-01-16)
Nitrogen-bisphosphonates (N-BPs) are the most widely used drugs for bone fragility disorders. Long-term or high-dose N-BP use is associated with unusual serious side effects such as osteonecrosis of the jaw, musculoskeletal pain, and atypical fractures of long bones. It has
M Mancuso et al.
Current drug targets, 11(1), 111-121 (2009-12-19)
Coenzyme Q10 (CoQ10, or ubiquinone) is an electron carrier of the mitochondrial respiratory chain (electron transport chain) with antioxidant properties. In view of the involvement of CoQ10 in oxidative phosphorylation and cellular antioxidant protection a deficiency in this quinone would
Meghan J Ho et al.
The Cochrane database of systematic reviews, (4)(4), CD007435-CD007435 (2009-10-13)
Studies have shown that coenzyme Q10 deficiency is associated with cardiovascular disease. Hypertension is a commonly measured surrogate marker for non-fatal and fatal cardiovascular endpoints such as heart attacks and strokes. Clinical trials have suggested that coenzyme Q10 supplementation can
Gerwyn Morris et al.
Molecular neurobiology, 48(3), 883-903 (2013-06-14)
Coenzyme Q10 (CoQ10) is an antioxidant, a membrane stabilizer, and a vital cofactor in the mitochondrial electron transport chain, enabling the generation of adenosine triphosphate. It additionally regulates gene expression and apoptosis; is an essential cofactor of uncoupling proteins; and
D Orsucci et al.
Current medicinal chemistry, 18(26), 4053-4064 (2011-08-10)
Coenzyme Q10 is a small electron carrier of the respiratory chain with antioxidant properties, widely used for the treatment of mitochondrial disorders. Mitochondrial diseases are neuromuscular disorders caused by impairment of the respiratory chain and increased generation of reactive oxygen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service