Merck
Skip To
P1049
Microcapillary tube, calibrated
capacity 50 μL
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Pack SizeSKUAvailabilityPrice
About This Item
NACRES:
NB.01
UNSPSC Code:
41121500
Manufacturer/tradename:
Drummond Scientific 2-000-050
Material:
glass
Packaging:
pack of 250 ea
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
glass
packaging
pack of 250 ea
manufacturer/tradename
Drummond Scientific 2-000-050
measuring range
accuracy: ±0.5%
capacity
50 μL
color
color-coded green
General description
  • Drummond Calibrated Micropipettes provide accuracy and convenience for a wide variety of liquid transfer applications.
  • Made of high quality glass.
  • Fire polished micropipettes are imprinted with volume and a color-code bar for easy identification.
  • Calibrated "To Deliver."
High quality Drummond micropipettes are fire polished, imprinted with volume and a color-code bar for easy identification. Calibrated "To Deliver."
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
P0799P0674P1174
manufacturer/tradename
Drummond Scientific 2-000-050
manufacturer/tradename
Drummond Scientific 2-000-020
manufacturer/tradename
Drummond Scientific 2-000-010
manufacturer/tradename
Drummond Scientific 2-000-100
material
glass
material
glass
material
glass
material
glass
packaging
pack of 250 ea
packaging
pack of 250 ea
packaging
pack of 250 ea
packaging
pack of 250 ea
capacity
50 μL
capacity
20 μL
capacity
10 μL
capacity
100 μL
color
color-coded green
color
color-coded black
color
color-coded orange
color
color-coded blue
measuring range
accuracy: ±0.5%
measuring range
accuracy: ±0.5%
measuring range
accuracy: ±0.5%
measuring range
accuracy: ±0.5%
Still not finding the right product?
Explore all of our products under Microcapillary tube, calibrated
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Global Trade Item Number
SKUGTIN
P1049-1PAK04061833566527