Skip to Content
Merck

Skip To

M0512

Methyl cellulose

viscosity: 4,000 cP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

wood (pulp cellulose)

Quality Level

form

powder

color

white to off-white

viscosity

3,500-5,600 cP, 2 % in water(20 °C)

solubility

water: 20 mg/mL, clear to hazy, colorless to light yellow

storage temp.

room temp

SMILES string

[*]OC[C@H]1O[C@@H](O[C@@H]2[C@@H](CO[*])O[C@@H](O[*])[C@H](O[*])[C@H]2O[*])[C@H](O[*])[C@@H](O[*])[C@@H]1O[*]

InChI key

YLGXILFCIXHCMC-JHGZEJCSSA-N

General description

Methyl cellulose is a water-soluble, non-ionic polymer with hydrophilic glucosidic bonds, derived from the polysaccharide cellulose.

Application

Methyl cellulose has been used:
  • in colony forming assays
  • for sperm motility assessment
  • for the generation of spheroids
  • in preparation of agar beads

Biochem/physiol Actions

Methyl cellulose mediates stabilization of oil, water and air mixtures, like foam, by emulsification. Methyl cellulose is used as a binder and thickener for various adhesives. It is also used as a treatment of constipation.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M038794378M0430
biological source

wood (pulp cellulose)

biological source

wood (pulp cellulose)

biological source

-

biological source

-

solubility

water: 20 mg/mL, clear to hazy, colorless to light yellow

solubility

-

solubility

-

solubility

-

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

form

powder

form

powder

form

powder

form

powder

storage temp.

room temp

storage temp.

room temp

storage temp.

-

storage temp.

-

color

white to off-white

color

white to off-white

color

-

color

-


Still not finding the right product?

Explore all of our products under Methyl cellulose


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Immunohistochemical screening for beta 6-integrin subunit expression in adenocarcinomas using a novel monoclonal antibody reveals strong up-regulation in pancreatic ductal adenocarcinomas in vivo and in vitro
Sipos B, et al.
Histopathology, 45(3), 226-236 (2004)
Reza Nosrati et al.
Clinical chemistry, 62(3), 458-465 (2016-01-10)
More than 70 million couples worldwide are affected by infertility, with male-factor infertility accounting for about half of the cases. Semen analysis is critical for determining male fertility potential, but conventional testing is costly and complex. Here, we demonstrate a
Cellulose and Cellulose Derivatives in the Food Industry: Fundamentals and Applications (2014)



Global Trade Item Number

SKUGTIN
M0512-500G04061833993279
M0512-100G04061838820563
M0512-1KG04061833993255
M0512-250G04061833993262

Questions

1–5 of 5 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. does it is suitable for chicken heterophil isolation?

    1 answer
    1. This product is typically used for applications involving carbohydrates and may not be specifically designed for chicken heterophil isolation. It is suggested to conduct a quick test to evaluate its effectiveness for this particular application.

      Helpful?

  3. Can I autoclave this product once solubilised?

    1 answer
    1. A 0.5-1% solution of the product in water can be autoclaved. However, at higher concentrations chunks of methyl cellulose could come out of solution. For that reason, autoclaving of those higher concentrations is not recommended.

      Helpful?

  4. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  5. What is the molecular weight of this methyl cellulose ?

    1 answer
    1. Based on the viscosity, this product has an expected molecular weight of 88,000 daltons. Please see the link below to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/251/785/m0512pis.pdf

      Helpful?

Reviews

No rating value

Active Filters