Skip to Content
Merck
All Photos(3)

Documents

M0430

Sigma-Aldrich

Methyl cellulose

meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

Assay

26.0-33.0% (methoxyl group, on Dry Basis)

form

powder

useful pH range

5.0-8.0 (2% in water)

viscosity

320-480 cP, 2 % in water(20 °C)

application(s)

pharmaceutical (small molecule)

SMILES string

[*]OC[C@H]1O[C@@H](O[C@@H]2[C@@H](CO[*])O[C@@H](O[*])[C@H](O[*])[C@H]2O[*])[C@H](O[*])[C@@H](O[*])[C@@H]1O[*]

InChI key

YLGXILFCIXHCMC-JHGZEJCSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl cellulose is an odorless and tasteless material. It is tough and non-toxic. Due to these properties, methyl cellulose is used as an adhesive in the food packaging industry. In water-soluble adhesives, it is utilized as a thickening agent.

Application

Methyl cellulose has been used as a supplement of Mammacult media for mammosphere culture from SUM159 TN breast cancer cells. It has also been used to formulate drugs such as ITI-007 and PCC0104005 to characterize their functional activity.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In Vitro and In Vivo Characterization of PCC0104005, a Novel Modulator of Serotonin-Dopamine Activity, as an Atypical Antipsychotic Drug
Xu Y,et al.
Scientific Reports, 8(1), 6892-6892 (2018)
Nuclear basic fibroblast growth factor regulates triple-negative breast cancer chemo-resistance
Li S, et al.
Breast Cancer Research, 17(1), 91-91 (2015)
Yanan Xu et al.
Scientific reports, 8(1), 6892-6892 (2018-05-04)
PCC0104005 is a novel drug candidate for treating schizophrenia that displays high affinity for serotonin, dopamine, and noradrenaline receptors, including partial agonism at dopamine D2, D3, D4, serotonin 5-HT1A, and 5-HT2A receptors and antagonism at 5-HT2B, 5-HT6, and 5-HT7 receptors.
Handbook of Adhesives and Surface Preparation (2011)
Functional profile of a novel modulator of serotonin, dopamine, and glutamate neurotransmission
Snyder GL, et al.
Psychopharmacology, 232(3), 605-621 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service