Skip to Content
Merck

Skip To

K4894

Poloxamer 188

solid

Synonym(s):

Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), Lutrol F68

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12161902
NACRES:
NA.75
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

agency

USP/NF, meets USP testing specifications

form

solid

mol wt

8.400 g/mol

pH

5.0-7.5(2.5% solution)

application(s)

sample preservation

SMILES string

O2C(C2)C.O1CC1

InChI

1S/C3H6O.C2H4O/c1-3-2-4-3;1-2-3-1/h3H,2H2,1H3;1-2H2

InChI key

RVGRUAULSDPKGF-UHFFFAOYSA-N

General description

Poloxamer 188 is a non-ionic amphiphilic copolymer, which has a central chain of hydrophobic polyoxypropylene and a hydrophilic polyoxyethylene at the ends.[1]

Application

Poloxamer 188 has been used as a supplement in F17 medium for the culture of HEK293-6E cells. It has also been used as a component of flow cytometry (FACS) buffer as a non-ionic detergent that protects cells from hydrodynamic damage.
Poloxamer 188 has been used as a component of cell-dissociation/minimal essential media (MEM) to sort tumor cells using EpCAM antibody-conjugated magnetic beads.[2]

Biochem/physiol Actions

Poloxamer 188 exhibits cytoprotective, hemorheological, anti-inflammatory, antithrombotic activities. It maintains the stability of membrane barrier function. P188 lowers surface tension and hydrophobic-based cellular adherence. It also inhibits lipid peroxidation stimulated by Fe2+ and H2O2.[1]

Legal Information


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.37112P489416758
form

solid

form

solid

form

solid, waxy solid

form

powder

application(s)

sample preservation

application(s)

liquid formulation
ophthalmics
parenterals
pharma/biopharma processes
pharmaceutical
semi-solid formulation
solid formulation
solubility enhancement

application(s)

-

application(s)

-

mol wt

8.400 g/mol

mol wt

-

mol wt

-

mol wt

-

Quality Level

300

Quality Level

500

Quality Level

100

Quality Level

100

pH

5.0-7.5(2.5% solution)

pH

5-7.5 (20 °C, 10 g/L in H2O)

pH

5.0-7.5

pH

5.0-7.5

agency

USP/NF, meets USP testing specifications

agency

ChP, NF, Ph. Eur.

agency

-

agency

-


Still not finding the right product?

Explore all of our products under Poloxamer 188


Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Antibody humanization by molecular dynamics simulations?in-silico guided selection of critical backmutations
Christian Margretter
Journal of Molecular Recognition (2016)
S Dhara et al.
Nature communications, 12(1), 3044-3044 (2021-05-26)
Unlike other malignancies, therapeutic options in pancreatic ductal adenocarcinoma (PDAC) are largely limited to cytotoxic chemotherapy without the benefit of molecular markers predicting response. Here we report tumor-cell-intrinsic chromatin accessibility patterns of treatment-naïve surgically resected PDAC tumors that were subsequently
S Dhara et al.
Nature communications, 12(1), 3044-3044 (2021-05-26)
Unlike other malignancies, therapeutic options in pancreatic ductal adenocarcinoma (PDAC) are largely limited to cytotoxic chemotherapy without the benefit of molecular markers predicting response. Here we report tumor-cell-intrinsic chromatin accessibility patterns of treatment-naïve surgically resected PDAC tumors that were subsequently



Global Trade Item Number

SKUGTIN
K4894-500G04061835568765

Questions

  1. How long can a 0,001% solution be stored at 4°C?

    1 answer
    1. The stability of a 0.001% solution at 4°C has not been tested. A solution of 100 mg/ml in water is stable for 1 year at -80°C and for 1 month at -20°C. It is recommended to aliquot the stock solutions to avoid repeated freeze-thaw cycles.

      Helpful?

Reviews

No rating value

Active Filters