Skip to Content
Merck

Skip To

16758

Poloxamer 407

purified, non-ionic

Synonym(s):

Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352104
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

form

powder

quality

purified

impurities

≤1.0% water

pH

5.0-7.5

solubility

soluble, clear, colorless

cation traces

Al: ≤5 mg/kg, As: ≤0.05 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤50 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤1000 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤50 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

UV absorption

λ: 260 nm Amax: ≤0.3, λ: 280 nm Amax: ≤0..2

suitability

no residue for filter test

storage temp.

2-8°C

SMILES string

O2C(C2)C.O1CC1

InChI

1S/C3H6O.C2H4O/c1-3-2-4-3;1-2-3-1/h3H,2H2,1H3;1-2H2

InChI key

RVGRUAULSDPKGF-UHFFFAOYSA-N

General description

Poloxamer 407 is a water-soluble, non-ionic surfactant with low toxicity and good solubilizing capacity. This triblock copolymer is composed of a hydrophobic residue of polyoxypropylene (POP) flanked by two hydrophilic units of polyoxyethylene (POE).

Application

Poloxamer 407 has been used:
  • as an inhibitor of lipoprotein lipase activity to assess hepatic very-low-density lipoprotein (VLDL)- triglyceride (TG) secretion in mice
  • for VLDL secretion assay in mice
  • to intraperitoneally inject the mice by dissolving in phosphate buffer saline (PBS) for hepatic triglyceride production

Biochem/physiol Actions

Poloxamer 407 (P407) exhibits reversible gelation properties above a particular polymer concentration and temperature. Poloxamer 407-based sterile injectable formulations serve to be a good candidate for drug delivery. P407 thermo-responsive hydrogels are broadly used as mucosal drug delivery systems as they do not irritate the mucosal membranes. P407 is called an “inactive ingredient” by the U.S. Food and Drug Administration (FDA) for several drug products like oral solutions, suspensions, and inhalation formulations.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P2164030620351.37112
suitability

no residue for filter test

suitability

-

suitability

-

suitability

-

form

powder

form

-

form

powder

form

solid

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

2-25°C

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

500

cation traces

Al: ≤5 mg/kg, Ba: ≤5 mg/kg, Ca: ≤50 mg/kg, Co: ≤5 mg/kg, Cu: ≤5 mg/kg, K: ≤1000 mg/kg, Mg: ≤5 mg/kg, Mo: ≤5 mg/kg, Ni: ≤5 mg/kg, Sr: ≤5 mg/kg, As: ≤0.05 mg/kg, Cd: ≤5 mg/kg, Fe: ≤5 mg/kg, Mn: ≤5 mg/kg, Pb: ≤5 mg/kg, Bi: ≤5 mg/kg, Li: ≤5 mg/kg, Zn: ≤5 mg/kg, Cr: ≤5 mg/kg, Na: ≤50 mg/kg

cation traces

-

cation traces

-

cation traces

-

impurities

≤1.0% water

impurities

-

impurities

≤0.25% ethylene glycol and diethylene glycol, ≤0.4% total ash, ≤0.75% water, ≤1 ppm ethylene oxide, ≤2 ppm arsenic traces, ≤20 ppm heavy metals, ≤5 ppm dioxan, ≤5 ppm propylene oxide, 50-125 ppm butylhydroxytoluene

impurities

-


Still not finding the right product?

Explore all of our products under Poloxamer 407


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Nae-Gyune Rim et al.
ACS biomaterials science & engineering, 3(8), 1542-1556 (2017-10-03)
Silk is a promising material for biomedical applications, and much research is focused on how application-specific, mechanical properties of silk can be designed synthetically through proper amino acid sequences and processing parameters. This protocol describes an iterative process between research
Camila S Freitas et al.
Parasitology research, 120(1), 321-335 (2020-11-17)
Treatment for visceral leishmaniasis (VL) is hampered mainly by drug toxicity, their high cost, and parasite resistance. Drug development is a long and pricey process, and therefore, drug repositioning may be an alternative worth pursuing. Cardenolides are used to treat
Hua Qing et al.
Cell, 182(2), 372-387 (2020-07-02)
Acute psychological stress has long been known to decrease host fitness to inflammation in a wide variety of diseases, but how this occurs is incompletely understood. Using mouse models, we show that interleukin-6 (IL-6) is the dominant cytokine inducible upon



Global Trade Item Number

SKUGTIN
I6758-1G04061835367078
I6758-10G04061835367061
I6758-5G04061835367085
420290-5GM04055977210828
420322-10GM07790788055622
420291-1GM04055977210835
420291-5GM07790788050108
420322-500GM07790788050153
70527-307790788052690
420322-100GM07790788050146
420322-1GM04055977210552
420322-25GM07790788057817
420322-5GM04055977210620
16758-250G04061838749628
I1284-5ML04061833847640
71300-307790788053185
71300-404055977253191
70235-404055977258585
69450-304055977274332
70837-304055977272574
70836-304055977272550

Questions

1–4 of 4 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What does the number in poloxamer-407, 105, 108, etc. stand for?

    1 answer
    1. The numeric suffix in various forms of Poloxamers refers to the mass per mole of polyoxypropylene and the percentage of polyoxyethylene present in that specific compound.

      Helpful?

  4. In poloxamer-407,105, 108 etc. This number stands for?

    1 answer
    1. Poloxamers are block co-polymers that have a basic structure of one central polyoxypropylene core adjoined to two polyoxyethylene side blocks or chains. The numeric suffix designated in various forms of Poloxamers is in reference to the mass per mole of polyoxypropylene and the percentage of polyoxyethylene present within that specific compound.
      See the link below for additional information. Examples can be found in Table 1 on page 2.

      https://nmr.oxinst.com/assets/uploads/MagRes/App%20Notes/X-Pulse/App_Note_3_Poloxamer_Oct_2019.pdf

      Helpful?

Reviews

No rating value

Active Filters