Skip to Content
Merck

Skip To

I0160

Imipenem monohydrate

≥98% (HPLC), B-lactam antibiotic, powder

Synonym(s):

(5R,6S)-6-[(1R)-1-Hydroxyethyl]-3-[[2-[(iminomethyl)amino]ethyl]thio]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid monohydrate, Primaxin monohydrate, Tienam monohydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C12H17N3O4S · H2O
CAS Number:
Molecular Weight:
317.36
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
264-734-5
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Imipenem monohydrate, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

antibiotic activity spectrum

mycobacteria

mode of action

cell wall synthesis | interferes

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

storage temp.

−20°C

SMILES string

O.C[C@@H](O)[C@@H]1[C@H]2CC(SCCNC=N)=C(N2C1=O)C(O)=O

InChI

1S/C12H17N3O4S.H2O/c1-6(16)9-7-4-8(20-3-2-14-5-13)10(12(18)19)15(7)11(9)17;/h5-7,9,16H,2-4H2,1H3,(H2,13,14)(H,18,19);1H2/t6-,7-,9-;/m1./s1

InChI key

GSOSVVULSKVSLQ-JJVRHELESA-N

General description

Chemical structure: β-lactam

Application

Imipenem monohydrate has been used to determine the resistance profiles of Pseudomonas aeruginosa isolates. It has also been used in susceptibility testing against Acinetobacter baumannii.

Biochem/physiol Actions

Imipenem is found effective against gram positive and negative aerobes and anaerobes. Imipenem is often combined with cilastatin, to inhibit its metabolism in kidney.
Imipenem monohydrate is a broad spectrum B-lactam antibiotic.
Imipenem monohydrate is a broad spectrum B-lactam antibiotic. It is a member of the carbapenem class of "magic bullet" antibiotics for severe infections.

Features and Benefits

This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR17961337809SML0037
assay

≥98% (HPLC)

assay

-

assay

-

assay

90-102% anhydrous basis (HPLC)

form

powder

form

solid

form

-

form

crystalline powder

Quality Level

100

Quality Level

300

Quality Level

-

Quality Level

100

storage temp.

−20°C

storage temp.

-10 to -25°C

storage temp.

−20°C

storage temp.

2-8°C

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

originator

-

originator

-

originator

-

mode of action

cell wall synthesis | interferes

mode of action

-

mode of action

-

mode of action

cell wall synthesis | interferes


Still not finding the right product?

Explore all of our products under Imipenem monohydrate


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters