Skip to Content
Merck
All Photos(1)

Key Documents

SML0037

Sigma-Aldrich

Ceftibuten hydrate

≥98% (HPLC)

Synonym(s):

(6R,7R)-7-[[(2Z)-2-(2-Amino-4-thiazolyl)-4-carboxy-1-oxo-2-buten-1-yl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14N4O6S2 · xH2O
CAS Number:
Molecular Weight:
410.42 (anhydrous basis)
MDL number:
UNSPSC Code:
51284115
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: ≥50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

O.Nc1nc(cs1)\C(=C\CC(O)=O)C(=O)N[C@H]2[C@H]3SCC=C(N3C2=O)C(O)=O

InChI

1S/C15H14N4O6S2.H2O/c16-15-17-7(5-27-15)6(1-2-9(20)21)11(22)18-10-12(23)19-8(14(24)25)3-4-26-13(10)19;/h1,3,5,10,13H,2,4H2,(H2,16,17)(H,18,22)(H,20,21)(H,24,25);1H2/b6-1-;/t10-,13-;/m1./s1

InChI key

JMLVKZRNRBPPDZ-TXOBIQCCSA-N

Related Categories

General description

Chemical structure: ß-lactam

Biochem/physiol Actions

Ceftibuten is a β-lactam antibiotic with antibacterial activity. It is effective for mild-to-moderate respiratory infections. It displays high stability against extended-spectrum β-lactamases producing bacteria.
Ceftibuten is a third generation cephalosporin antibiotic
Third generation cephalosporin antibiotic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Staffan Mårild et al.
Pediatric nephrology (Berlin, Germany), 24(3), 521-526 (2008-09-27)
A randomized, open, coordinated multi-center trial compared the bacteriological and clinical efficacy and safety of orally administered ceftibuten and trimethoprim-sulfamethoxazole (TMP-SMX) in children with febrile urinary tract infection (UTI). Children aged 1 month to 12 years presenting with presumptive first-time
D Adam et al.
Fortschritte der Medizin. Originalien, 119 Suppl 2, 63-70 (2005-02-12)
Group A Streptococci have remained sensitive to penicillins and other betalactam antibiotics, e. g. cephalosporins. Since the beginning of the 1950s oral penicillin V given three times daily in a dose of 50,000 IU daily has been the drug of
Kouji Kimura et al.
Journal of clinical microbiology, 47(12), 4154-4157 (2009-10-09)
Although group B streptococcus (GBS) has been considered to be uniformly susceptible to beta-lactams, the presence of GBS with reduced penicillin susceptibility (PRGBS) was recently confirmed genetically. We developed a feasible and reliable method for screening PRGBS in clinical microbiology
Megumi Irie et al.
Pflugers Archiv : European journal of physiology, 449(2), 186-194 (2004-09-02)
Small peptides and some pharmacologically active compounds are absorbed from the small intestine by the apical H(+)-coupled peptide transporter 1 (PEPT1) and the basolateral peptide transporter. Here we investigated the efflux properties of the basolateral peptide transporter in Caco-2 cells
Burkholderia cepacia complex nasal isolation in immunocompetent patients with sinonasal polyposis not associated with cystic fibrosis.
G Marioni et al.
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology, 26(1), 73-75 (2006-12-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service