Skip to Content
Merck

Skip To

87128

Tetracycline

98.0-102.0% (HPLC)

Synonym(s):

Tetracycline HCL, Tetracycline Monohydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C22H24N2O8 · xH2O
CAS Number:
Molecular Weight:
444.43 (anhydrous basis)
UNSPSC Code:
51284017
NACRES:
NA.85
PubChem Substance ID:
EC Number:
200-481-9
Beilstein/REAXYS Number:
2230417
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

assay

98.0-102.0% (HPLC)

form

powder or crystals

color

faint yellow to dark yellow

mp

172-174 °C (dec.)

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O)C(=O)c4c(O)cccc4[C@@]3(C)O

InChI

1S/C22H24N2O8/c1-21(31)8-5-4-6-11(25)12(8)16(26)13-9(21)7-10-15(24(2)3)17(27)14(20(23)30)19(29)22(10,32)18(13)28/h4-6,9-10,15,25,27-28,31-32H,7H2,1-3H3,(H2,23,30)/t9-,10-,15-,21+,22-/m0/s1

InChI key

OFVLGDICTFRJMM-WESIUVDSSA-N

Gene Information

General description

Chemical structure: tetracycline

Biochem/physiol Actions

Mode of Action: Tetracycline passively diffuses through proin channels in the cell membrane, binding to 30S ribosomes and inhibits protein synthesis by preventing access of aminoacyl tRNA to the acceptor site on the mRNA-ribosome complex. It also binds to the bacterial 50S ribosomal subunit, altering the membrane and causing intracellular components to leak from bacterial cells. The inhibitory effects can be reversed by washing, suggesting that it is the reversibly bound antibiotic, and not the irreversibly bound drug, that is responsible for antibacterial action.

Mode of Resistance: The effects are inactivated via a loss of cell wall permeability.

Antimicrobial spectrum: Includes a wide range of antimicrobial activity against gram-positive and gram-negative bacteria.

Packaging

25g, 100g

Other Notes

Keep container tightly closed in a dry and well-ventilated place.Hygroscopic. Light sensitive. Keep in a dry place.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T3258T8032T3383
antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder or crystals

form

powder or crystals

form

powder

form

powder

assay

98.0-102.0% (HPLC)

assay

98.0-102.0% (HPLC)

assay

-

assay

≥95% (European Pharmacopoeia HPLC assay)

biological source

synthetic

biological source

-

biological source

synthetic

biological source

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?

Explore all of our products under Tetracycline


signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
87128-25G04061835515677
87128-100G04061835541836

Questions

1–4 of 4 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How to prepare stock solutions for item 87128-25G, Tetracycline?

    1 answer
    1. Tetracycline is soluble in methanol, giving a clear solution at up to 30 mg/ml. It is practically insoluble in chloroform and ether, very slightly soluble in water, slightly soluble in alcohol, very soluble in dilute hydrochloric acid and alkali hydroxide solutions.

      Helpful?

  4. devo fare lo stock della tetraciclina. come e in cosa la devo diluire?

    1 answer
    1. This product is soluble in methanol or ethanol, giving a completely clear solution at up to 30 mg/ml. The stability of this material in solution is pH-dependent. Stock solutions prepared in ethanol are estimated to be stable for 1-3 months when aliquoted and stored at -20°C. Please see the link below to review the product datasheet for T3258, an identical tetracycline product of another brand. The information provided applies to both products.
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/406/734/t3258dat.pdf

      Helpful?

Reviews

No rating value

Active Filters